Skip to Content
Merck
All Photos(2)

Documents

163333

Sigma-Aldrich

Geraniol

98%

Synonym(s):

trans-3,7-Dimethyl-2,6-octadien-1-ol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C=CHCH2CH2C(CH3)=CHCH2OH
CAS Number:
Molecular Weight:
154.25
Beilstein:
1722456
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

5.31 (vs air)

vapor pressure

~0.2 mmHg ( 20 °C)

Assay

98%

form

liquid

refractive index

n20/D 1.474 (lit.)

bp

229-230 °C (lit.)

solubility

water: soluble 0.1 g/L at 25 °C

density

0.879 g/mL at 20 °C (lit.)

SMILES string

C\C(C)=C\CC\C(C)=C\CO

InChI

1S/C10H18O/c1-9(2)5-4-6-10(3)7-8-11/h5,7,11H,4,6,8H2,1-3H3/b10-7+

InChI key

GLZPCOQZEFWAFX-JXMROGBWSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Geraniol undergoes oxidation with molecular oxygen in supercritical carbon dioxide catalyzed by chromium containing mesoporous molecular sieve to yield citral.

Application

Geraniol was used in field evaluation of synthetic herbivore-induced plant volatiles as attractants to beneficial insects. It was used to evaluate the tumor-suppressive potency of isoprenoids in vitro and in vivo.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

226.4 °F - closed cup

Flash Point(C)

108 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ying Zhou et al.
Biomolecules, 9(12) (2019-12-06)
When insects attack plants, insect-derived elicitors and mechanical damage induce the formation and emission of plant volatiles that have important ecological functions and flavor properties. These events have mainly been studied in model plants, rather than crop plants. Our study
L He et al.
The Journal of nutrition, 127(5), 668-674 (1997-05-01)
Sundry mevalonate-derived constituents (isoprenoids) of fruits, vegetables and cereal grains suppress the growth of tumors. This study estimated the concentrations of structurally diverse isoprenoids required to inhibit the increase in a population of murine B16(F10) melanoma cells during a 48-h
Selective catalytic oxidation of geraniol to citral with molecular oxygen in supercritical carbon dioxide.
Dapurkar SE, et al.
Applied Catalysis A: General, 394(1), 209-214 (2011)
Maria Cristina Bonferoni et al.
Pharmaceutics, 11(3) (2019-03-06)
The pharmacological activities of geraniol include anticancer and neuroprotective properties. However, its insolubility in water easily induces separation from aqueous formulations, causing administration difficulties. Here we propose new emulsified formulations of geraniol by using the amphiphilic polymer chitosan-oleate (CS-OA) as
Arunachalam Kannappan et al.
Journal of medical microbiology, 66(10), 1506-1515 (2017-09-13)
The current study has been designed to delineate the efficacy of geraniol (GE) on biofilm formation in Staphylococcus epidermidis as well as the effect of subinhibitory concentrations of GE on the development of adaptive resistance. Biofilm biomass quantification assay was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service