Skip to Content
Merck
All Photos(2)

Key Documents

E22201

Sigma-Aldrich

Ethyl diazoacetate

contains ≥13 wt. % dichloromethane

Synonym(s):

DAAE

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
N=N=CHCOOC2H5
CAS Number:
Molecular Weight:
114.10
Beilstein:
107654
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form

liquid

Quality Level

contains

≥13 wt. % dichloromethane

refractive index

n20/D 1.46 (lit.)

bp

140-141 °C/720 mmHg (lit.)

mp

−22 °C (lit.)

solubility

water: slightly soluble

density

1.085 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CCOC(=O)C=[N+]=[N-]

InChI

1S/C4H6N2O2/c1-2-8-4(7)3-6-5/h3H,2H2,1H3

InChI key

YVPJCJLMRRTDMQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reagent for ruthenium-catalyzed asymmetric cyclopropanation of alkenes.

related product

Product No.
Description
Pricing

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 3 - Self-react. E - Skin Irrit. 2

Storage Class Code

5.2 - Organic peroxides and self-reacting hazardous materials

WGK

WGK 3

Flash Point(F)

116.6 °F - closed cup

Flash Point(C)

47 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lian-Biao Zhao et al.
The Journal of organic chemistry, 72(26), 10276-10278 (2007-11-29)
Copper-catalyzed [4 + 1] cycloaddition reaction of alpha,beta-acetylenic ketones with alpha-diazo esters offers an efficient, direct route to highly substituted furans. The reaction conditions and the scope of the process are examined, and a possible mechanism is proposed.
Barry M Trost et al.
Journal of the American Chemical Society, 131(5), 1674-1675 (2009-02-05)
Magnesium-catalyzed enantioselective aldol between ethyl diazoacetate and aromatic, aliphatic, and alpha,beta-unsaturated aldehydes affords alpha-diazo-beta-hydroxy-esters in high enantioselectivities. Aldol adducts resulting from this asymmetric transformation are versatile intermediates toward the synthesis of several ester containing chiral building blocks.
Ying Chen et al.
The Journal of organic chemistry, 69(7), 2431-2435 (2004-03-31)
Vitamin B(12) derivatives were found for the first time to be general and efficient catalysts for asymmetric cyclopropanation of alkenes with ethyl diazoacetate (EDA). Among several common derivatives, aquocobalamin (B(12a)) was shown to be the most effective catalyst for a
Marianne Lenes Rosenberg et al.
The Journal of organic chemistry, 76(8), 2465-2470 (2011-03-19)
The performance of recently reported highly cis-diastereoselective Rh(I) cyclopropanation catalysts has been significantly improved by a systematic study of different reaction parameters (catalyst activation, solvent, temperature, stoichiometry). The catalyst efficiency and diastereoselectivity were enhanced by changing the activating agent from
Yoichiro Kuninobu et al.
Chemical communications (Cambridge, England), 46(46), 8860-8862 (2010-10-23)
Treatment of tertiary amines with diazoacetate in the presence of a catalytic amount of an iron salt, FeCl(3), in ethanol gave glycine derivatives. In this reaction, a carbon-nitrogen single bond of the amine was cleaved.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service