DWK241701701
DURAN® reaction flask with ground joint
glass flask, flask capacity (50 mL), single neck, round bottom, flask joint: ST/NS 19/26
Synonym(s):
241701701
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
Recommended Products
material
glass flask
round bottom
Joint
joint: ST/NS flask19/26
feature
lid
packaging
case of 10 ea
manufacturer/tradename
DURAN® 241701701
flask capacity
(50 mL)
Looking for similar products? Visit Product Comparison Guide
General description
Tight closure due to standard ground joint. The geometry permits very uniform heating. Closable with glass and plastic stopper
Legal Information
DURAN is a registered trademark of DWK Life Sciences
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service