Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
DURAN
®
reaction flask with ground joint
Products
Product Category
flasks (21)
Capacity (mL)
Package Size
Filter Products
Showing 1-20 of 21
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
DWK241702706
DURAN
®
reaction flask with ground joint,
glass flask, flask capacity (100 mL), single neck, round bottom, neck joint: ST/NS 29/32
Expand
View Pricing
DWK241703608
DURAN
®
reaction flask with ground joint,
glass flask, flask capacity (250 mL), single neck, round bottom frit, neck joint: ST/NS 24/29
Expand
View Pricing
DWK241703702
DURAN
®
reaction flask with ground joint,
glass flask, flask capacity (250 mL), single neck, round bottom, neck joint: ST/NS 29/32
Expand
View Pricing
DWK241705403
DURAN
®
reaction flask with ground joint,
glass flask, flask capacity (1,000 mL), single neck, round bottom, neck joint: ST/NS 24/29
Expand
View Pricing
DWK241704604
DURAN
®
reaction flask with ground joint,
glass flask, flask capacity (500 mL), single neck, round bottom, neck joint: ST/NS 29/32
Expand
View Pricing
DWK241702509
DURAN
®
reaction flask with ground joint,
glass flask, flask capacity (100 mL), single neck, round bottom, neck joint: ST/NS 14/23
Expand
View Pricing
DWK241702603
DURAN
®
reaction flask with ground joint,
glass flask, flask capacity (100 mL), single neck, round bottom, neck joint: ST/NS 24/29
Expand
View Pricing
DWK241701804
DURAN
®
reaction flask with ground joint,
glass flask, flask capacity (50 mL), single neck, round bottom, neck thread joint: ST/NS 24/29
Expand
View Pricing
DWK241705703
DURAN
®
reaction flask with ground joint,
glass flask, flask capacity (1,000 mL), single neck, round bottom, neck joint: ST/NS 45/40
Expand
View Pricing
DWK241701701
DURAN
®
reaction flask with ground joint,
glass flask, flask capacity (50 mL), single neck, round bottom, flask joint: ST/NS 19/26
Expand
View Pricing
DWK241702003
DURAN
®
reaction flask with ground joint,
glass flask, flask capacity (50 mL), single neck, round bottom, neck joint: ST/NS 14/23
Expand
View Pricing
DWK241702406
DURAN
®
reaction flask with ground joint,
flask capacity (100 mL), glass flask, single neck, round bottom, neck joint: ST/NS 19/26
Expand
View Pricing
DWK241706305
DURAN
®
reaction flask with ground joint,
glass flask, flask capacity (2,000 mL), single neck, round bottom, neck joint: ST/NS 29/32
Expand
View Pricing
DWK241701307
DURAN
®
reaction flask with ground joint,
glass flask, flask capacity (25 mL), single neck, round bottom, neck joint: ST/NS 14/23
Expand
View Pricing
DWK241704707
DURAN
®
reaction flask with ground joint,
glass flask, flask capacity (500 mL), single neck, round bottom, neck joint: ST/NS 45/40
Expand
View Pricing
DWK241705609
DURAN
®
reaction flask with ground joint,
glass flask, flask capacity (1,000 mL), single neck, round bottom, neck joint: ST/NS 29/32
Expand
View Pricing
DWK241701401
DURAN
®
reaction flask with ground joint,
glass flask, flask capacity (25 mL), single neck, round bottom, neck joint: ST/NS 19/26
Expand
View Pricing
DWK241707207
DURAN
®
reaction flask with ground joint,
glass flask, flask capacity (4,000 mL), single neck, round bottom, neck joint: ST/NS 45/40
Expand
View Pricing
DWK241701907
DURAN
®
reaction flask with ground joint,
glass flask, flask capacity (50 mL), single neck, round bottom, neck joint: ST/NS 29/32
Expand
View Pricing
DWK241704407
DURAN
®
reaction flask with ground joint,
glass flask, flask capacity (500 mL), single neck, round bottom frit, neck joint: ST/NS 24/29
Expand
View Pricing
Page 1 of 2
Page 1 of 2