All Photos(2)
About This Item
Linear Formula:
BrCH2CH(Br)CH2OH
CAS Number:
Molecular Weight:
217.89
Beilstein:
1719127
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
98%
form
liquid
refractive index
n20/D 1.559 (lit.)
bp
95-97 °C/10 mmHg (lit.)
density
2.12 g/mL at 25 °C (lit.)
SMILES string
OCC(Br)CBr
InChI
1S/C3H6Br2O/c4-1-3(5)2-6/h3,6H,1-2H2
InChI key
QWVCIORZLNBIIC-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Synthetic building block, fire-proofing agent, and carcinogen.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 1B - Repr. 2
Storage Class Code
6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK
WGK 3
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
R Benigni
Mutagenesis, 6(5), 423-425 (1991-09-01)
A QSAR model based on the combination of two molecular descriptors--estimated electrophilic reactivity and Ashby's structural alerts--was used to predict the carcinogenicity of 44 chemicals currently bioassayed by the US National Toxicology Program. These predictions will be compared with the
Ijaz Gul et al.
Analytical biochemistry, 609, 113971-113971 (2020-09-27)
Enzyme immobilization using inorganic membranes has enticed increased attention as they not only improve enzyme stability, but also furnish user-friendly biodevices that can be tailored to different applications. Herein, we explored the suitability of the glass fiber membrane for enzyme
G K Hemakanthi De Alwis et al.
Archives of environmental contamination and toxicology, 53(1), 134-139 (2007-03-31)
2,3-Dibromo-1-propanol (DBP) was used as an active flame retardant in the 1970s. It was also used as an intermediate in the preparation of insecticide formulations, pharmaceuticals and the flame retardants tris(2,3-dibromopropyl) phosphate (Tris-BP) and tetrabromobisphenol A bis (2,3-dibromopropyl ether). DBP
G Choudhary
American Industrial Hygiene Association journal, 48(9), 809-813 (1987-09-01)
One of the starting materials for the preparation of TRIS-BP (tris-2,3-dibromopropanol phosphate), which is used in the treatment of fabrics and various materials to make them nonflammable, is 2,3-dibromopropanol. Because of the reported toxic nature of its parent phosphate compound
2,3-Dibromopropan-1-ol.
IARC monographs on the evaluation of carcinogenic risks to humans, 77, 439-453 (2000-12-02)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service