Skip to Content
Merck
All Photos(1)

Key Documents

36295

Sigma-Aldrich

2,3-Dichloro-1-propanol

≥97.0% (GC)

Synonym(s):

Glycerol-α,β-dichlorohydrin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2CHClCH2OH
CAS Number:
Molecular Weight:
128.99
Beilstein:
1732060
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥97.0% (GC)

density

1.360 g/mL at 20 °C (lit.)

functional group

chloro
hydroxyl

SMILES string

OCC(Cl)CCl

InChI

1S/C3H6Cl2O/c4-1-3(5)2-6/h3,6H,1-2H2

InChI key

ZXCYIJGIGSDJQQ-UHFFFAOYSA-N

General description

2,3-Dichloro-1-propanol belongs to the group of chloropropanols. Inhibitory effects of 2,3-dichloro-1-propanol on T cell both in vivo and in vitro is reported. Improved enantioselective resolution of (R,S)-2,3-dichloro-1-propanol to (S)-2, 3-dichloro-1-propanol by whole cells of a recombinant Escherichia coli in n-heptane-aqueous biphasic system is reported.

Application

2,3-Dichloro-1-propanol may be employed as carbon and energy supplement for the growth of Pseudomonas putida strain (MC4).

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 3 Oral - Eye Irrit. 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

199.4 °F

Flash Point(C)

93 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yoshiki Kuroda et al.
Journal of UOEH, 24(3), 271-280 (2002-09-19)
A rare outbreak of acute hepatic damage in workers exposed to dichloropropanols was reported in 1992. As there are no detailed reports of dichloropropanols (DCPs) toxicity and its mechanism, we reviewed the toxicity of dichloropropanols using our results. 1) A
Shu-Ping Zou et al.
Journal of biotechnology, 188, 42-47 (2014-08-26)
The enantioselective resolution of (R,S)-2,3-dichloro-1-propanol ((R,S)-DCP) to (S)-DCP by whole cells of a recombinant Escherichia coli expressing halohydrin dehalogenase (HHDH) activity was limited by product inhibition. To solve this problem to improve the productivity of (S)-DCP, an n-heptane-aqueous biphasic system
Ijaz Gul et al.
Biotechnology and applied biochemistry, 67(4), 685-692 (2020-02-18)
Epoxides are widely used chemicals, the determination of which is of paramount importance. Herein, we present an enzyme-based approach for noninstrumental detection of epoxides in standard solution and environmental samples. Halohydrin dehalogenase (HheC) as a biological recognition element and epichlorohydrin
K Imazu et al.
Journal of UOEH, 14(1), 73-76 (1992-03-01)
We investigated the toxicity of dichloropropanols (DCPs) in hematological findings and serum chemistry. The solutions of two isomers of DCPs, 1,3-dichloro-2-propanol (DC2P) and 2,3-dichloro-1-propanol (DC1P) were dissolved in saline at the concentration of 100 mg/ml, and 0.1 ml of each
Jing Lu et al.
International immunopharmacology, 17(2), 321-328 (2013-07-16)
2,3-Dichloro-1-propanol (2,3-DCP) is a member of a group of chemicals known as chloropropanols. Currently, immunotoxicity of 2,3-DCP has not been reported. In the present study, we studied its inhibitory effects on T cell both in vivo and in vitro. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service