D221201
Tetradecanedioic acid
99%
Synonym(s):
Dodecanedicarboxylic acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
HOOC(CH2)12COOH
CAS Number:
Molecular Weight:
258.35
Beilstein:
1788202
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
99%
form
powder
mp
124-127 °C (lit.)
SMILES string
OC(=O)CCCCCCCCCCCCC(O)=O
InChI
1S/C14H26O4/c15-13(16)11-9-7-5-3-1-2-4-6-8-10-12-14(17)18/h1-12H2,(H,15,16)(H,17,18)
InChI key
HQHCYKULIHKCEB-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
I Ziegler et al.
Blut, 44(4), 231-240 (1982-04-01)
Using the Crithidia assay 3.0 ng biopterin/ml blood was found, of which one third was present in the plasma. The erythrocyte fraction comprised 1.7 ng and the buffy coat 0.33 ng. After Ficoll separation 0.050 ng were found in the
Julien Herrou et al.
The Journal of biological chemistry, 293(19), 7437-7456 (2018-03-24)
The general stress response sigma factor σE1 directly and indirectly regulates the transcription of dozens of genes that influence stress survival and host infection in the zoonotic pathogen Brucella abortus Characterizing the functions of σE1-regulated genes therefore would contribute to
Dolors Company-Arumí et al.
Phytochemical analysis : PCA, 27(6), 326-335 (2016-10-22)
Protective plant lipophilic barriers such as suberin and cutin, with their associated waxes, are complex fatty acyl derived polyesters. Their precise chemical composition is valuable to understand the specific role of each compound to the physiological function of the barrier.
Chie Ohdoi et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 792(1), 123-130 (2003-06-28)
Lesch-Nyhan syndrome (LNS) is caused by a severe deficiency of hypoxanthine-guanine phosphoribosyltransferase (HPRT) and clinically characterized by self-injurious behavior and nephrolithiasis; the latter is treatable with allopurinol, an inhibitor of xanthine oxidase which converts xanthine and hypoxanthine into uric acid.
Ling Li et al.
Molecular cell, 80(3), 525-540 (2020-10-18)
Well-balanced and timed metabolism is essential for making a high-quality egg. However, the metabolic framework that supports oocyte development remains poorly understood. Here, we obtained the temporal metabolome profiles of mouse oocytes during in vivo maturation by isolating large number of
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service