Skip to Content
Merck
All Photos(1)

Documents

177482

Sigma-Aldrich

1,16-Hexadecanediol

97%

Synonym(s):

Hexadecamethylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO(CH2)16OH
CAS Number:
Molecular Weight:
258.44
Beilstein:
1811931
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

bp

197-199 °C/3 mmHg (lit.)

mp

91-94 °C (lit.)

SMILES string

OCCCCCCCCCCCCCCCCO

InChI

1S/C16H34O2/c17-15-13-11-9-7-5-3-1-2-4-6-8-10-12-14-16-18/h17-18H,1-16H2

InChI key

GJBXIPOYHVMPQJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

1,16-Hexadecanediol was used to activate Mps1 MAP kinase pathway. It was also used to accumulate BChl c monoesterified with the corresponding diols.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Misa Kuroki et al.
Scientific reports, 7(1), 9697-9697 (2017-08-31)
The rice blast fungus Magnaporthe oryzae differentiates a specialized infection structure called an appressorium to invade rice cells. In this report, we show that CBP1, which encodes a chitin-deacetylase, is involved in the induction phase of appressorium differentiation. We demonstrate
Takashi Fujikawa et al.
Molecular microbiology, 73(4), 553-570 (2009-07-16)
Oligosaccharides derived from cell wall of fungal pathogens induce host primary immune responses. To understand fungal strategies circumventing the host plant immune responses, cell wall polysaccharide localization was investigated using fluorescent labels during infectious structure differentiation in the rice blast
Risato Nishimori et al.
Biochemistry, 50(36), 7756-7764 (2011-08-19)
Unnatural bacteriochlorophyll (BChl) c derivatives possessing a hydroxy group at the terminus of a hydrocarbon chain at the 17-propionate were biosynthesized in the green sulfur photosynthetic bacterium Chlorobaculum tepidum. Addition of exogenous 1,8-octanediol, 1,12-dodecanediol, and 1,16-hexadecanediol in acetone to liquid
Ying Li et al.
mBio, 11(2) (2020-03-27)
The appressoria that are generated by the rice blast fungus Magnaporthe oryzae in response to surface cues are important for successful colonization. Previous work showed that regulators of G-protein signaling (RGS) and RGS-like proteins play critical roles in appressorium formation.
Martin Korn et al.
PloS one, 10(5), e0125960-e0125960 (2015-05-21)
We used insertional mutagenesis by Agrobacterium tumefaciens mediated transformation (ATMT) to isolate pathogenicity mutants of Colletotrichum higginsianum. From a collection of 7200 insertion mutants we isolated 75 mutants with reduced symptoms. 19 of these were affected in host penetration, while

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service