Skip to Content
Merck
All Photos(1)

Key Documents

D186007

Sigma-Aldrich

2,2-Dimethylsuccinic acid

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOOCCH2C(CH3)2COOH
CAS Number:
Molecular Weight:
146.14
Beilstein:
1764476
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
crystals
Assay:
99%

Quality Level

Assay

99%

form

crystals

mp

138-142 °C (lit.)

SMILES string

CC(C)(CC(O)=O)C(O)=O

InChI

1S/C6H10O4/c1-6(2,5(9)10)3-4(7)8/h3H2,1-2H3,(H,7,8)(H,9,10)

InChI key

GOHPTLYPQCTZSE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

María C Bernini et al.
Inorganic chemistry, 49(11), 5063-5071 (2010-05-01)
Two new layered polymeric frameworks have been synthesized under different hydrothermal conditions and characterized by single-crystal X-ray diffraction, thermal analysis, and variable temperature-Fourier Transform Infrared Spectroscopy (VT-FTIR). The compound I, with formula [Er(2)(dms)(3)(H(2)O)(4)], has a triclinic cell with parameters a
G Giordano et al.
Rapid communications in mass spectrometry : RCM, 4(5), 170-172 (1990-05-01)
2-(2'-octenyl)succinic acid has been identified in urine samples from children investigated for a possible inherited metabolic disease. Its structural identification has been achieved by gas chromatography/mass spectrometry using both electron ionization and chemical ionization and by tandem mass spectrometry (MS/MS)
Wen-Jun Song
Talanta, 170, 74-80 (2017-05-16)
A new fluorescent sensing platform based on ultrathin metal-organic framework (MOF) nanosheets (MnDMS) was prepared from the flexible ligand 2,2-dimethylsuccinate and Mn ions. The MnDMS nanoparticles can be obtained by simply ultrasonication of the MnDMS crystal, and then can be
Birgitta C Burckhardt et al.
Journal of the American Society of Nephrology : JASN, 13(11), 2628-2638 (2002-10-25)
The active transport of Krebs cycle intermediates, such as succinate, alpha-ketoglutarate, and citrate, is mediated by sodium-coupled transporters found in the luminal (NaDC-1) and basolateral plasma membranes (NaDC-3) of proximal tubule cells. This study used the two-electrode voltage clamp technique
R I Kelley
Pediatric research, 30(6), 564-569 (1991-12-01)
The excretion of 2-(2'-octenyl)succinic acid (OSA) and several metabolites of OSA was studied by gas chromatography/mass spectrometry in 17 infants and children fed one of three proprietary elemental or protein-hydrolysate formulas that use OSA-modified cornstarch as an emulsifying agent. Variable

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service