Skip to Content
Merck
All Photos(1)

Key Documents

D4379

Sigma-Aldrich

3,3-Dimethylglutaric acid

98%

Synonym(s):

3,3-Dimethylpentanedioic acid, NSC 14987, NSC 49114

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOOCCH2C(CH3)2CH2COOH
CAS Number:
Molecular Weight:
160.17
Beilstein:
1765439
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

powder

mp

100-102 °C (lit.)

SMILES string

CC(C)(CC(O)=O)CC(O)=O

InChI

1S/C7H12O4/c1-7(2,3-5(8)9)4-6(10)11/h3-4H2,1-2H3,(H,8,9)(H,10,11)

InChI key

DUHQIGLHYXLKAE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant involved in:
  • Cyclodehydration of diols
  • Synthesis of conjugates of betulin derivatives used as anti-HIV agents
  • Preparation of dimeric peptide antagonists of IgG-FcRn interaction
  • Microwave-assisted protection of glutaraldehyde
  • Synthesis of glycyrrhetinic acid derivatives for proteasome inhibition
  • Catalytic, asymmetric transannular aldolizations

Quality

May contain trace amounts of toluene.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shiou-Shiow Farn et al.
Applied radiation and isotopes : including data, instrumentation and methods for use in agriculture, industry and medicine, 127, 61-67 (2017-05-20)
[
Colleen O'Connor et al.
Journal of biomedical materials research. Part A, 105(5), 1243-1251 (2017-02-09)
A family of pH-responsive terpolymers composed of methacrylic acid (MAA), N-vinyl pyrrolidone (NVP), and poly(ethylene glycol) monomethylether monomethacrylate (PEGMMA) have been developed and evaluated for their pH-responsive swelling behavior, protein-loading capabilities, and cytocompatibility. These terpolymer hydrogels, designated as P((MAA-co-NVP)-g-EG), were
Elaine L Ferguson et al.
Journal of drug targeting, 25(9-10), 818-828 (2017-07-22)
'Polymer Enzyme Liposome Therapy' (PELT) is a two-step anticancer approach in which a liposomal drug and polymer-phospholipase conjugate are administered sequentially to target the tumour interstitium by the enhanced permeability and retention effect, and trigger rapid, local, drug release. To
Kevin A Kovalchik et al.
Rapid communications in mass spectrometry : RCM, 31(24), 2057-2065 (2017-09-26)
The characterization of naphthenic acid fraction compounds (NAFCs) in oil sands process affected water (OSPW) is of interest for both toxicology studies and regulatory reasons. Previous studies utilizing authentic standards have identified dicarboxylic naphthenic acids using two-dimensional gas chromatography hyphenated
D J Cosgrove et al.
Journal of experimental botany, 45(Spec Iss), 1711-1719 (1994-11-01)
Walls isolated from cucumber hypocotyls retain autolytic activities and the ability to extend when placed under the appropriate conditions. To test whether autolysis and extension are related, we treated the walls in various ways to enhance or inhibit long-term wall

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service