D182257
3,5-Dimethyl-1-pyrazolylformaminidium nitrate
97%
Synonym(s):
1-Amidino-3,5-dimethylpyrazole nitrate, 3,5-Dimethylpyrazole-1-carboxamidine nitrate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C6H10N4 · HNO3
CAS Number:
Molecular Weight:
201.18
Beilstein:
3599170
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
97%
form
powder
mp
167-168 °C (dec.) (lit.)
SMILES string
O[N+]([O-])=O.Cc1cc(C)n(n1)C(N)=N
InChI
1S/C6H10N4.HNO3/c1-4-3-5(2)10(9-4)6(7)8;2-1(3)4/h3H,1-2H3,(H3,7,8);(H,2,3,4)
InChI key
AGYXIUAGBLMBGV-UHFFFAOYSA-N
Application
Guanylating reagent for amines.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Orakan Hanpanich et al.
Nucleosides, nucleotides & nucleic acids, 39(1-3), 156-169 (2019-10-15)
The cationic copolymer poly(L-lysine)-graft-dextran (PLL-g-Dex) has nucleic acid chaperone-like activity. The copolymer facilitates both DNA hybridization and strand exchange reactions. For these reasons, DNA-based enzyme (DNAzyme) activity is enhanced in the presence of copolymer. In this study, we evaluated activities
Journal of Carbohydrate Chemistry, 12, 1173-1173 (1993)
Peter Pogány et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 71(4), 1466-1473 (2008-06-20)
In the paper a joint experimental and theoretical study of 3,5-dimethyl-1H-pyrazole-1-carboxamidine (L) as well as its complexes CoL2(H2O)2(NO3)2 and NiL2(H2O)2(NO3)2 is reported. On the basis of FT-IR experiments and a DFT-derived scaled quantum mechanical force field the normal coordinate analysis
A Bernhardt et al.
The journal of peptide research : official journal of the American Peptide Society, 50(2), 143-152 (1997-08-01)
Peptide-4-nitroanilides can be quickly synthesised using an Fmoc-based approach on 2-chlorotritylchloride resin. Preformed building blocks Fmoc-Xaa-NH-Np (Xaa = Cit, Cys, Gln, His, Lys, Orn, Ser, Thr, Tyr, Trp) can be attached via side chain to the 2-chlorotritylchloride linker of the
L Spero et al.
Biochimica et biophysica acta, 251(3), 345-356 (1971-12-28)
Guanidination of the free amino groups of staphylococcal enterotoxin B with 3,5-dimethyl-1-guanylpyrazole converted 31-32 of 33 epsilon-amino groups and 30% of the N-terminal residue. This product, although markedly reduced in solubility, suffered no gross change in conformation and retained full
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service