Skip to Content
Merck
All Photos(2)

Documents

D174904

Sigma-Aldrich

2,6-Dimethylphenol

≥99.5%

Synonym(s):

2-Hydroxy-m-xylene, vic.-m-Xylenol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C6H3OH
CAS Number:
Molecular Weight:
122.16
Beilstein:
1446677
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99.5%

form

crystals

autoignition temp.

1110 °F

bp

203 °C (lit.)

mp

43-45 °C (lit.)

SMILES string

Cc1cccc(C)c1O

InChI

1S/C8H10O/c1-6-4-3-5-7(2)8(6)9/h3-5,9H,1-2H3

InChI key

NXXYKOUNUYWIHA-UHFFFAOYSA-N

Gene Information

human ... GABRA1(2554)

Looking for similar products? Visit Product Comparison Guide

Related Categories

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

186.8 °F - closed cup

Flash Point(C)

86 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gertrud Haeseler et al.
British journal of pharmacology, 145(7), 916-925 (2005-05-25)
Phenol derivatives constitute a family of neuroactive compounds. The aim of our study was to identify structural features that determine their modulatory effects at glycine receptors. We investigated the effects of four methylated phenol derivatives and two halogenated analogues on
Tingting Li et al.
Talanta, 78(4-5), 1497-1502 (2009-04-14)
In the present work, a simple, selective, and sensitive method has been proposed for the determination of four phenols (catechol, resorcinol, 2,6-dimethylphenol, and 2,4,6-trinitrophenol) in water samples. The method is based on poly-(methacrylic acid-co-ethylene glycol dimethacrylate) monolith microextraction (PMME) and
O Grech-Bélanger et al.
Research communications in chemical pathology and pharmacology, 58(1), 53-62 (1987-10-01)
A new metabolite, 2,6-dimethylphenol obtained by O-dealkylation of mexiletine by rabbit liver was identified. A g.c.-f.i.d. method was developed for its analysis in hepatic homogenates and some of the characteristics of the metabolic reaction were studied. Formation of 2,6-dimethylphenol is
J Hartung et al.
Zentralblatt fur Bakteriologie, Mikrobiologie und Hygiene. 1. Abt. Originale B, Hygiene, 179(5), 431-439 (1984-10-01)
Dust-borne phenols and indols were extracted by ethanol from the sedimentation dust of two pig houses (finishing pigs on half slatted floor resp. piglets on deep litter) and a hen house (3-stage battery cages) and analysed by gas chromatography. In
H Yang et al.
Journal of magnetic resonance. Series B, 105(3), 205-210 (1994-11-01)
Modified Jeener solid-echo pulse sequences are proposed for the measurement of the proton dipolar spin-lattice relaxation time, T1D, of motionally restricted (solid-like) components in the presence of mobile molecular species, such as encountered in biological tissue. A phase-cycled composite-pulse sequence

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service