Skip to Content
Merck
All Photos(3)

Key Documents

D11582

Sigma-Aldrich

1,2-Diaminoanthraquinone

Synonym(s):

1,2-Diamino-9,10-anthraquinone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H10N2O2
CAS Number:
Molecular Weight:
238.24
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form

powder

Quality Level

mp

289-291 °C (lit.)

SMILES string

Nc1ccc2C(=O)c3ccccc3C(=O)c2c1N

InChI

1S/C14H10N2O2/c15-10-6-5-9-11(12(10)16)14(18)8-4-2-1-3-7(8)13(9)17/h1-6H,15-16H2

InChI key

LRMDXTVKVHKWEK-UHFFFAOYSA-N

Application

1,2-Diaminoanthraquinone (DAQ) can be used as a building block to prepare:
  • Imidazoanthraquinone derivatives via palladium-catalyzed Suzuki–Miyaura and Sonogashira coupling reactions.
  • Anthra[1,2-d]imidazole-6,11-diones by reacting with various aldehydes in the presence of phosphotungstic acid/PEG-400 catalyst.
  • DAQ-containing polymer materials are applicable as solid-state colorimetric sensors for nitrite anion and nitric oxide.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cross-linked poly (2-hydroxyethylmethacrylate) films doped with 1, 2-diaminoanthraquinone (DAQ) as efficient materials for the colorimetric sensing of nitric oxide and nitrite anion
Bru Miriam, et al.
Tetrahedron Letters, 47(11), 1787-1791 (2006)
Francisco Galindo et al.
Photochemical & photobiological sciences : Official journal of the European Photochemistry Association and the European Society for Photobiology, 7(1), 126-130 (2008-01-03)
Spectroscopic analysis of the fluorescent probe 1,2-diaminoanthraquinone () provides information about the mechanism of nitric oxide imaging in living cells. Fluorescent aggregates of a reaction product of are thought to be responsible for the images obtained with confocal fluorescence microscopy.
P Heiduschka et al.
Neuroreport, 9(18), 4051-4057 (1999-02-02)
The role of nitric oxide (NO) as a putative mediator of neuronal death can be understood best if NO is detected directly. For this purpose, we developed a new sensitive method that for the first time directly captures released NO
[Detection of NO-synthase activity of lactobacilli by fluorescent staining].
D R Iarullina et al.
Mikrobiologiia, 76(4), 570-572 (2007-11-03)
María J Marín et al.
Chembiochem : a European journal of chemical biology, 12(16), 2471-2477 (2011-09-29)
Nitric oxide (NO) is involved in many biological processes. Aromatic ortho-diamine derivatives are commonly used in the fluorescence imaging of NO in living cells. ortho-diamino (o-diamino) compounds are believed to react with NO in an oxygenated medium leading to the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service