Skip to Content
Merck
All Photos(1)

Key Documents

CDS000624

Sigma-Aldrich

1-Amino-2-butanol

AldrichCPR

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H11NO
CAS Number:
Molecular Weight:
89.14
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

solid

SMILES string

CCC(O)CN

InChI

1S/C4H11NO/c1-2-4(6)3-5/h4,6H,2-3,5H2,1H3

InChI key

KODLUXHSIZOKTG-UHFFFAOYSA-N

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stephanie M Smith et al.
Journal of biomedical materials research. Part B, Applied biomaterials, 108(5), 1915-1924 (2019-12-28)
Interactions between contact lens multipurpose solution (MPS) components and the contact lenses with which they are used are both lens and solution dependent. As such, lens dimensional changes may arise after cleaning and immersion cycling in different lens care solutions
Marc A Masen et al.
PloS one, 15(9), e0239363-e0239363 (2020-09-25)
Healthcare workers around the world are experiencing skin injury due to the extended use of personal protective equipment (PPE) during the COVID-19 pandemic. These injuries are the result of high shear stresses acting on the skin, caused by friction with
Radu-Liviu Sumalan et al.
Molecules (Basel, Switzerland), 25(7) (2020-04-08)
The discovery of environmentally friendly and inexpensive plant growth regulators (PGRs) for agronomically important crops is a necessity and must be considered a priority worldwide. This study provides the synthesis, structure determination and the biological evaluation of two binary organic
B Nozière et al.
Physical chemistry chemical physics : PCCP, 20(3), 1583-1590 (2017-12-21)
Naturally-occurring inorganic ammonium ions have been recently reported as efficient catalysts for some organic reactions in water, which contributes to the understanding of the chemistry in some natural environments (soils, seawater, atmospheric aerosols, …) and biological systems, and is also

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service