Skip to Content
Merck
All Photos(1)

Documents

14960

Sigma-Aldrich

Bis(2-hydroxypropyl)amine

≥98.0% (T)

Synonym(s):

1,1′-Iminodi-2-propanol, Diisopropanolamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH[CH2CH(OH)CH3]2
CAS Number:
Molecular Weight:
133.19
Beilstein:
605363
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0% (T)

form

solid

impurities

≤1% water

bp

249-250 °C/745 mmHg (lit.)

mp

42-45 °C (lit.)

solubility

H2O: miscible
alcohol: miscible

density

1.004 g/mL at 25 °C (lit.)

SMILES string

CC(O)CNCC(C)O

InChI

1S/C6H15NO2/c1-5(8)3-7-4-6(2)9/h5-9H,3-4H2,1-2H3

InChI key

LVTYICIALWPMFW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Bis(2-hydroxypropyl)amine (Diisopropanolamine) was used to study its effects upon choline uptake and phospholipid synthesis in Chinese hamster ovary (CHO) cells.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

275.0 °F - closed cup

Flash Point(C)

135 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L M Gieg et al.
Canadian journal of microbiology, 45(5), 377-388 (1999-08-14)
Diisopropanolamine (DIPA) is a "sweetening agent" used to remove hydrogen sulfide from sour natural gas, and it is a contaminant at some sour gas treatment facilities in western Canada. To investigate the biodegradation of this alkanolamine, 14C-DIPA was used in
K A Johnson et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 45(10), 1838-1845 (2007-05-18)
The repeated dose oral and dermal toxicity of diisopropanolamine (DIPA) was evaluated in rats and compared to the reported toxicity of the related secondary alcohol amine, diethanolamine (DEA). Fischer 344/DuCrl rats were given up to 750 mg/kg/day by dermal application
W T Stott et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 46(2), 761-766 (2007-11-09)
Aminoalcohols differ in mammalian toxicity at least in part based upon their ability to alter the metabolism of phospholipids and to cause depletion of the essential nutrient choline in animals. This study examined the incorporation of diisopropanolamine (DIPA) into phospholipids
Contact dermatitis due to diisopropanolamine.
K Fujimoto et al.
Contact dermatitis, 21(1), 56-56 (1989-07-01)
Two cases of contact dermatitis due to diisopropanolamine.
Yoshihiro Umebayashi
The Journal of dermatology, 32(2), 145-146 (2005-05-24)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service