All Photos(1)
About This Item
Linear Formula:
BrC10H6OH
CAS Number:
Molecular Weight:
223.07
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
97%
form
powder
mp
78-81 °C (lit.)
SMILES string
Oc1ccc2ccccc2c1Br
InChI
1S/C10H7BrO/c11-10-8-4-2-1-3-7(8)5-6-9(10)12/h1-6,12H
InChI key
FQJZPYXGPYJJIH-UHFFFAOYSA-N
Related Categories
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Chaoren Shen et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 23(54), 13369-13378 (2017-06-27)
The palladium-catalyzed chemoselective carbonylation of bromoaryl triflates is reported. The selective C-Br bond versus C-OTf (OTf=triflate) bond functionalization can be remarkably tuned by the combination of the ligand [4,5-bis(diphenylphosphino)-9,9-dimethylxanthene (Xantphos) vs. 1,1'-bis(diphenylphosphino)ferrocene (DPPF)] and the solvent (toluene vs. DMSO). The
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service