Skip to Content
Merck
All Photos(2)

Documents

B19401

Sigma-Aldrich

Benzyl cyanide

98%

Synonym(s):

Phenylacetonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2CN
CAS Number:
Molecular Weight:
117.15
Beilstein:
385941
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

0.1 mmHg ( 20 °C)

Assay

98%

form

liquid

refractive index

n20/D 1.523 (lit.)

bp

233-234 °C (lit.)

mp

−24 °C (lit.)

density

1.015 g/mL at 25 °C (lit.)

SMILES string

N#CCc1ccccc1

InChI

1S/C8H7N/c9-7-6-8-4-2-1-3-5-8/h1-5H,6H2

InChI key

SUSQOBVLVYHIEX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

215.6 °F - closed cup

Flash Point(C)

102 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Magzoub O Bashir et al.
Journal of insect physiology, 56(6), 640-645 (2010-02-09)
Previous studies had demonstrated stage differentiation in the cohesion (aggregation) pheromone systems of the desert locust, Schistocerca gregaria. In laboratory arena, the nymphal and adult stages responded aggregatively to their own pheromone, but dispersed evenly within the arena in the
Dunming Zhu et al.
Journal of biotechnology, 133(3), 327-333 (2007-12-07)
A nitrilase gene blr3397 from Bradyrhizobium japonicum USDA110 was cloned and over-expressed in Escherichia coli, and the encoded protein was purified to give a nitrilase with a single band of about 34.5kD on SDS-PAGE. The molecular weight of the holoenzyme
Hui-Lei Hou et al.
The Journal of organic chemistry, 77(5), 2553-2558 (2012-02-10)
Aerobic oxidations of dianionic C(60) were examined in PhCN and PhCH(2)CN, where dioxygen was activated to O(2)(•-) via the single-electron transfer from C(60)(2-) and underwent oxygenation and dehydrogenation reactions, respectively. Addition of PhCH(2)Br led to further benzylation for the oxygenated
Tekla Strzalko et al.
The Journal of organic chemistry, 77(15), 6431-6442 (2012-06-30)
Mechanisms of alkylation by PhCH(2)Cl or CH(3)I in THF and of deuteriation by DCl (4 N in D(2)O) in THF or THF-toluene of lithiated phenylacetonitrile monoanions and dianions obtained with LHMDS, LDA, or n-BuLi are studied by vibrational and NMR
Jacques Corset et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 68(5), 1340-1346 (2007-05-01)
The carbanionic species possibly formed during the first alkylation step of phenylacetonitrile lithiated anion by CH(3)I and PhCH(2)Cl in THF-hexane solution and their complexes with the lithium salt formed have been observed by infrared spectrometry and characterized by density functional

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service