B15805
Benzyl acetate
≥99%
Synonym(s):
Acetic acid benzyl ester
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Linear Formula:
CH3COOCH2C6H5
CAS Number:
Molecular Weight:
150.17
Beilstein:
1908121
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
vapor pressure
23 mmHg ( 110 °C)
Assay
≥99%
form
liquid
autoignition temp.
862 °F
refractive index
n20/D 1.502 (lit.)
bp
206 °C (lit.)
mp
−51 °C (lit.)
density
1.054 g/mL at 25 °C (lit.)
SMILES string
CC(=O)OCc1ccccc1
InChI
1S/C9H10O2/c1-8(10)11-7-9-5-3-2-4-6-9/h2-6H,7H2,1H3
InChI key
QUKGYYKBILRGFE-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 3
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
203.0 °F - closed cup
Flash Point(C)
95 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
C Debonneville et al.
Analytical chemistry, 74(10), 2345-2351 (2002-06-01)
A multisniffing system has been developed to allow three panelists to simultaneously participate in a GC-olfactometric analysis. This device, associated with a computerized data treatment, allows shortening CHARM and GC-"SNIF' analyses to less than 1 week and less than 1
Eşref Demir et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 48(5), 1239-1242 (2010-02-23)
In this study, different concentrations of four benzyl derivatives (benzyl alcohol, benzyl acetate, benzoic acid and benzaldehyde) used as flavour ingredients were investigated for genotoxicity in in vitro. By taking blood from two healthy people comet assay was carried on
G Y Wittman et al.
Journal of chromatography. A, 874(2), 225-234 (2000-05-19)
The direct derivatisation of acetic acid with n-hexyl chloroformate and with benzyl bromide in water was evaluated. With n-hexyl chloroformate, acetic acid did not give the n-hexyl acetate derivative, but the reaction of acetic acid with benzyl bromide in aqueous
J Caldwell
Teratogenesis, carcinogenesis, and mutagenesis, 13(4), 185-190 (1993-01-01)
The mouse lymphoma assay is commonly included in batteries of tests for the mutagenic and carcinogenic potential of chemicals. Unlike other bacterial and mammalian cell tests, it has an unacceptably high incidence of false positive results. This paper reviews data
Sotaro Momosaki et al.
Nuclear medicine and biology, 34(8), 939-944 (2007-11-14)
In order to develop a suitable radiotracer for the measurement of glial metabolism, we synthesized four different types of ester derivatives of [14C]acetate, namely, [14C]phenyl acetate, [14C]para-nitrophenyl acetate, [14C]2,4-dinitrophenyl acetate and [14C]benzyl acetate ([14C]BA), and evaluated their potencies in rats.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service