Skip to Content
Merck
All Photos(1)

Documents

W254800

Sigma-Aldrich

Heptyl alcohol

≥97%, FCC, FG

Synonym(s):

1-Heptanol, Heptyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)6OH
CAS Number:
Molecular Weight:
116.20
FEMA Number:
2548
Beilstein:
1731686
EC Number:
Council of Europe no.:
70
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
2.021
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 117
FDA 21 CFR 172.515

vapor pressure

0.5 mmHg ( 20 °C)

Assay

≥97%

autoignition temp.

662 °F

refractive index

n20/D 1.424 (lit.)

bp

176 °C (lit.)

mp

−36 °C (lit.)

density

0.822 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

apple; green; leafy; musty; fruity; vegetable; waxy

SMILES string

CCCCCCCO

InChI

1S/C7H16O/c1-2-3-4-5-6-7-8/h8H,2-7H2,1H3

InChI key

BBMCTIGTTCKYKF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Taste at 1-10 ppm

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

168.8 °F - closed cup

Flash Point(C)

76 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Klaus Neef et al.
The Journal of thoracic and cardiovascular surgery, 144(5), 1176-1184 (2012-09-18)
The effect of mechanical preconditioning on skeletal myoblasts in engineered tissue constructs was investigated to resolve issues associated with conduction block between skeletal myoblast cells and cardiomyocytes. Murine skeletal myoblasts were used to generate engineered tissue constructs with or without
Nannan Ye et al.
Lab on a chip, 11(6), 1096-1101 (2011-02-05)
Transport across gap junction channels (GJCs) between neighboring cells is critical to synchronizing cell's electrical and metabolic activities and maintaining cell homeostasis. Here we present a non-invasive microfluidic method to measure molecular diffusion across GJCs in multiple 1D cell arrays
Mathieu C Rémond et al.
American journal of physiology. Heart and circulatory physiology, 301(5), H1952-H1964 (2011-09-13)
This study examined transgenic mice whose expression of a β-galactosidase (lacZ) reporter is driven by a GATA6 gene enhancer. Previous investigations established that transcription of the transgene was associated with precardiac mesoderm and primary heart tube myocardium, which decreased progressively
Isaac K Kiplagat et al.
Electrophoresis, 32(21), 3008-3015 (2011-10-18)
Electromembrane extraction (EME) and CE with capacitively coupled contactless conductivity detection (CE-C(4) D) was applied to rapid and sensitive determination of perchlorate in drinking water and environmental samples. Porous polypropylene hollow fiber impregnated with 1-heptanol acted as a supported liquid
Gabriela Oliveira et al.
International journal of pharmaceutics, 393(1-2), 61-67 (2010-04-08)
While it is well recognised that formulation components influence drug permeation, few studies have addressed the influence of vehicles on drug transport in artificial or biological membranes Previously we have investigated the effects of temperature on the uptake of model

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service