Skip to Content
Merck
All Photos(1)

Key Documents

93498

Sigma-Aldrich

Glycerol trivalerate

≥98.0% (GC)

Synonym(s):

1,2,3-Trivalerylglycerol, Trivalerin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)3COOCH2]2CHOCO(CH2)3CH3
CAS Number:
Molecular Weight:
344.44
Beilstein:
1804221
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Pricing and availability is not currently available.

Quality Level

Assay

≥98.0% (GC)

form

liquid

refractive index

n20/D 1.439

density

1.002 g/mL at 20 °C (lit.)

functional group

ester

SMILES string

CCCCC(=O)OCC(COC(=O)CCCC)OC(=O)CCCC

InChI

1S/C18H32O6/c1-4-7-10-16(19)22-13-15(24-18(21)12-9-6-3)14-23-17(20)11-8-5-2/h15H,4-14H2,1-3H3

InChI key

PZJLFHDNXGAZHU-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

  • Choose from one of the most recent versions:

    Certificates of Analysis (COA)

    Lot/Batch Number

    Don't see the Right Version?

    If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service