Merck
Sensors design based on hybrid gold-silica nanostructures.
Biosensors & bioelectronics (2020-02-15)
Elnaz Bagheri, Legha Ansari, Elham Sameiyan, Khalil Abnous, Seyed Mohammad Taghdisi, Mohammad Ramezani, Mona Alibolandi
ABSTRACT
Hybrid silica-gold based sensors show attractive performance in sensing technologies. Due to their interesting optical properties and biological compatibility, gold nanoparticles (AuNPs) have been extensively implemented in sensing technology. Hybridization of AuNPs with silica NPs as a material with unique characteristic comprising large surface area, narrow pore distribution, tunable pore size and excellent charge transport provides great opportunity to fabricate promising sensing materials. This review summarizes the current developments on sensing devices based on gold-silica hybrid materials and discussing their interest in designing biosensors for improved analytes detection.
Sigma-Aldrich
Gold coated silica nanoparticles, 1 wt. % in water, 400 nm (non-porous silica), 20 nm (gold nanoparticles)
Sigma-Aldrich
Gold coated silica nanoparticles, 1 wt. % in water, 600 nm (non-porous silica), 20 nm (gold nanoparticles)
Sigma-Aldrich
Gold coated silica nanoparticles, 1 wt. % in water, 800 nm (non porous silica), 20 nm (gold nanoparticles)
Sigma-Aldrich
Gold coated silica nanoparticles, 1 wt. % in water, 200 nm (non-porous silica), 20 nm (gold nanoparticles)