Skip to Content
Merck
All Photos(3)

Key Documents

905348

Sigma-Aldrich

Rh2(R-p-Ph-TPCP)4

Synonym(s):

Dirhodium tetrakis((S,R)-1-(4-phenyl(phenyl))-2,2-diphenylcyclopropanecarboxylate), Dirhodium tetrakis[(R)-1-(4-phenyl(phenyl))-2,2- diphenylcyclopropane carboxylate], Tetrakis[(R)-1-(4-phenyl(phenyl))-2,2- diphenylcyclopropane carboxylato]dirhodium(II)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C112H84O8Rh2
CAS Number:
Molecular Weight:
1763.67
UNSPSC Code:
12352101
NACRES:
NA.22

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Changming Qin et al.
Journal of the American Chemical Society, 135(39), 14516-14519 (2013-09-13)
Rhodium-catalyzed reaction of vinyldiazoacetates with nitrones results in a formal [3+2]-cycloaddition to generate 2,5-dihydroisoxazoles with high levels of asymmetric induction. The cascade reaction begins with a vinylogous addition event, followed by an iminium addition ring-closure/hydride migration/alkene isomerization cascade. Dirhodium tetrakis(triarylcyclopropanecarboxylates)
Changming Qin et al.
Journal of the American Chemical Society, 136(27), 9792-9796 (2014-06-17)
The influence of sterically demanding dirhodium tetracarboxylate catalysts on the site selectivity of C-H functionalization by means of rhodium carbene-induced C-H insertion is described. The established dirhodium tetraprolinate-catalyzed reactions of aryldiazoacetates cause preferential C-H functionalization of secondary C-H bonds as
Liangbing Fu et al.
Journal of the American Chemical Society, 138(18), 5761-5764 (2016-04-12)
Enantioselective C-H functionalization of relatively electron-deficient methyl sites was achieved with the combination of 2,2,2-trichloroethyl aryldiazoacetates and tetrakis(triarylcyclopropanecarboxylate) dirhodium catalysts. The substrate scope of the transformation was relatively broad, and C-H functionalization products were furnished with excellent levels of enantioselectivity.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service