Skip to Content
Merck
All Photos(3)

Key Documents

905259

Sigma-Aldrich

Rh2(S-PTAD)4

Synonym(s):

Tetrakis[(S)-(+)-(1-adamantyl)-(N-phthalimido)acetato]dirhodium(II)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C80H80N4O16Rh2
CAS Number:
Molecular Weight:
1559.32
MDL number:
UNSPSC Code:
12161600
NACRES:
NA.22

form

powder or crystals

mp

>300 °C

SMILES string

[Rh]([Rh](OC(=O)[C@@H](N%19C(=O)c%20c(cccc%20)C%19=O)C%17%16C[C@@H]%18C[C@@H](C[C@@H](C%18)C%17)C%16)OC(=O)[C@@H](N%14C(=O)c%15c(cccc%15)C%14=O)C%12%11C[C@@H]%13C[C@@H](C[C@@H](C%13)C%12)C%11)(OC(=O)[C@@H](N9C(=O)c%10c(cccc%10)C9=O)C76C[C@@H]8C[C@@H](C[C@

InChI

1S/4C20H21NO4.2Rh/c4*22-17-14-3-1-2-4-15(14)18(23)21(17)16(19(24)25)20-8-11-5-12(9-20)7-13(6-11)10-20;;/h4*1-4,11-13,16H,5-10H2,(H,24,25);;/q;;;;2*+2/p-4/t4*11-,12+,13-,16-,20?;;/m1111../s1

InChI key

SGEDWOHAUXKUGM-PAPHCAFZSA-J

Application

This chiral Rh(II) dimer was developed in the Davies lab to perform asymmetric carbene and nitrene reactions (C-H insertion, cyclopropanation, aziridination, C-H amination) with high regio- and stereocontrol.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Justin R Denton et al.
Chemical communications (Cambridge, England), (10), 1238-1240 (2008-03-01)
Nitrile-substituted cyclopropanes are readily synthesized in a stereocontrolled fashion from the intermolecular cyclopropanation between 2-diazo-2-phenylacetonitrile and electron-rich olefins, catalyzed by the chiral dirhodium complex, Rh(2)(S-PTAD)(4).
Justin R Denton et al.
Organic letters, 11(4), 787-790 (2009-01-17)
The reaction of a variety of alpha-aryl-alpha-diazo ketones with activated olefins, catalyzed by the adamantyl glycine-derived dirhodium complex Rh(2)(S-PTAD)(4), generates cyclopropyl ketones with high diastereoselectivity (up to >95:5 dr) and enantioselectivity (up to 98% ee). Intermolecular C-H functionalization of 1,4-cyclohexadiene
Kuangbiao Liao et al.
Nature, 533(7602), 230-234 (2016-05-14)
The laboratory synthesis of complex organic molecules relies heavily on the introduction and manipulation of functional groups, such as carbon-oxygen or carbon-halogen bonds; carbon-hydrogen bonds are far less reactive and harder to functionalize selectively. The idea of C-H functionalization, in
Yan Su et al.
Organic letters, 18(17), 4356-4359 (2016-08-17)
Optically active cis-cyclopropane carboxylates are prepared via the Rh2(S-PTAD)4-catalyzed cyclopropanation of α-silyl styrenes with aryl diazoacetates followed by desilylation of the resulting silyl cyclopropane carboxylates. The conjugation of the aryl ring with C═C bond and π stacking are proposed for
Zhanjie Li et al.
Journal of the American Chemical Society, 132(1), 396-401 (2009-12-10)
The rhodium-catalyzed reaction of racemic allyl alcohols with methyl phenyldiazoacetate or methyl styryldiazoacetate results in a two-step process, an initial oxonium ylide formation followed by a [2,3]-sigmatropic rearrangement. This process competes favorably with the more conventional O-H insertion chemistry as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service