Skip to Content
Merck
All Photos(3)

Key Documents

901695

Sigma-Aldrich

Lithium tetrafluoroborate

≥98%, acid <200 ppm, anhydrous

Synonym(s):

Lithium borofluoride, Lithium fluoroborate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LiBF4
CAS Number:
Molecular Weight:
93.75
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.23

grade

anhydrous

Assay

≥98%

form

powder

mp

293-300 °C (dec.) (lit.)

application(s)

battery manufacturing

SMILES string

[Li+].F[B-](F)(F)F

InChI

1S/BF4.Li/c2-1(3,4)5;/q-1;+1

InChI key

UFXJWFBILHTTET-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Lithium tetrafluoroborate (LiBF4), a lithium salt, exhibits high conductivity, low electrical charge transfer, moisture resistance, and good cyclic properties. It is suitable for a low-temperature electrolytic system. Its performance is comparable to the widely used LiPF6 system.

Application

Lithium tetrafluoroborate (LiBF4) finds application as electrolyte material in lithium ion batteries (LIBs). However, the presence of trace amount of water in LiBF4 causes its hydrolysis which leads to HF generation and destruction of the battery electrodes. Our LiBF4 contains < 100 ppm of water and is well suited for applications in LIBs.

Caution

  • These electrolyte solutions have extremely low water content; Please handle under inert and moisture free environment (glove box).
  • Keep away from heat, sparks and flame. Avoid moisture, strong acid, strong alkalis, and oxidizing agents. Avoid skin and eye contact from product. Highly toxic and corrosive gases (HF) are generated by reaction with moisture os use only under inert gas atmosphere.
  • Please handle under inert and moisture free environment.

Storage:
Keep containers away from heat, humidity, direct sunlight and ignition source. Keep containers tightly closed in a cool dry and well-ventilated place. Do not store with strong oxidizing agents.

Legal Information

Product of Ube Industries Ltd.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Muta. 2 - Skin Corr. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vanchiappan Aravindan et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 17(51), 14326-14346 (2011-11-25)
This paper presents an overview of the various types of lithium salts used to conduct Li(+) ions in electrolyte solutions for lithium rechargeable batteries. More emphasis is paid towards lithium salts and their ionic conductivity in conventional solutions, solid-electrolyte interface
Investigation of the synergetic effects of LiBF 4 and LiODFB as wide-temperature electrolyte salts in lithium-ion batteries
Zhang Li
Ionics, 24(10), 2995-3004 (2018)
Conductivity and electrical properties of chitosan-methylcellulose blend biopolymer electrolyte incorporated with lithium tetrafluoroborate
Salman YAK, et al.
International Journal of Electrochemical Science, 13, 3185-3199 (2018)
Doi T, et al.
Journal of the Electrochemical Society, 163, A2211-A2215 (2016)
Zuo X, et al.
Journal of the Electrochemical Society, 160, A1199-A1204 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service