Skip to Content
Merck
All Photos(1)

Key Documents

729477

Sigma-Aldrich

Lithium-6Li tetrafluoroborate

95 atom % 6Li

Synonym(s):

Lithium-6Li fluoroborate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
6LiBF4
CAS Number:
Molecular Weight:
92.82
UNSPSC Code:
12352101
PubChem Substance ID:

isotopic purity

95 atom % 6Li

form

solid

mp

293-300 °C (dec) (lit.)

mass shift

M-1

SMILES string

[6Li][B](F)(F)(F)F

InChI

1S/BF4.Li/c2-1(3,4)5;/q-1;+1/i;1-1

InChI key

UFXJWFBILHTTET-HCMAANCNSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Muta. 2 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service