Skip to Content
Merck
All Photos(1)

Key Documents

86216

Sigma-Aldrich

Synperonic® PE P105

surfactant

Synonym(s):

Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), Poly(propylene glycol)-block-poly(ethylene glycol)-block-poly(propylene glycol)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

description

non-ionic

form

solid

impurities

≤1.0% water

mp

40-50 °C

hydroxyl value

15‑19 mg

SMILES string

O2C(C2)C.O1CC1

InChI

1S/C3H6O.C2H4O/c1-3-2-4-3;1-2-3-1/h3H,2H2,1H3;1-2H2

InChI key

RVGRUAULSDPKGF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Synperonic®PE P105 surfactant is a polyethylene oxide (PEO)-polypropylene oxide (PPO)-polyethylene oxide (PEO) based copolymer. It has two hydrophilic groups that are attached to a central hydrophobic group that self-assembles in aqueous solution into clusters known as micelles.

Legal Information

Synperonic is a registered trademark of Croda International PLC

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rheology of polyethylene oxide-polypropylene oxide block copolymer stabilized latices and emulsions
Faers MA and Luckham PF
Colloids and Surfaces. A, Physicochemical and Engineering Aspects, 86(21), 317-327 (1994)
An initial evaluation of Synperonic PE surfactants as soil washing agents
Paterson IF, et al.
Geological Society, London, Engineering Geology Special Publications, 14(1), 227-233 (1998)
Comparing colloidal phase separation induced by linear polymer and by microgel particles
Bayliss K, et al.
Soft Matter, 7(21), 10345-10352 (2011)
Burak Altun et al.
Ultrasonics, 110, 106274-106274 (2020-11-02)
Tissue-mimicking materials (TMMs) play a key role in the quality assurance of ultrasound diagnostic equipment and should have acoustic properties similar to human tissues. We propose a method to quantify the acoustic properties of TMM samples through the use of
Jianan Shen et al.
Biomaterials, 34(5), 1581-1590 (2012-11-14)
The metastasis of breast cancer is the leading cause of cancer death in women, and the lung is a common location of a secondary tumor that has metastasized from the primary source tumor. In this work, an attempt to simultaneously

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service