Skip to Content
Merck
All Photos(1)

Key Documents

857297

Sigma-Aldrich

D-(+)-Ribonic γ-lactone

97%

Synonym(s):

D(+)-Ribono-1,4-lactone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H8O5
CAS Number:
Molecular Weight:
148.11
Beilstein:
82057
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

crystals

optical activity

[α]24/D +18°, c = 1 in H2O

mp

85-87 °C (lit.)

SMILES string

OC[C@H]1OC(=O)[C@H](O)[C@@H]1O

InChI

1S/C5H8O5/c6-1-2-3(7)4(8)5(9)10-2/h2-4,6-8H,1H2/t2-,3-,4-/m1/s1

InChI key

CUOKHACJLGPRHD-BXXZVTAOSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Important building block for chiral acyclics, cyclopentenones, and oxabicyclic systems. Also employed in studies on nonlinear optical materials.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aldrichimica Acta, 22, 49-49 (1989)
Chemistry of Materials, 5, 802-802 (1993)
P C Raveendranath et al.
Carbohydrate research, 253, 207-223 (1994-02-03)
A series of 3-C-alkyl- (and 3-C-phenyl-) 2,3-dideoxy-D-erythro-pentono-1,4-lactones, compounds which are important in the synthesis of modified nucleosides and antibiotic sugars, were synthesized from D-ribonolactone. By a route that proceeded via 5-O-protected D-ribonolactone, 5-O-protected 2,3-dideoxy-D-glycero-pent-2-enono-1,4-lactones were synthesized and reacted with R2CuLi
Nucleosides. CXLVIII. Synthesis of 6-(beta-D-ribofuranosyl)picolinamide. A novel C-nucleoside from D-ribonolactone.
M M Kabat et al.
Chemical & pharmaceutical bulletin, 36(2), 634-640 (1988-02-01)
High resolution gas chromatographic/real-time high resolution mass spectrometric identification of organic acids in human urine.
S Lewis et al.
Analytical chemistry, 51(8), 1275-1285 (1979-07-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service