Skip to Content
Merck
All Photos(1)

Key Documents

310301

Sigma-Aldrich

L-Gulonic acid γ-lactone

95%

Synonym(s):

L-Gulonic γ-lactone, L-(+)-Gulono-1,4-lactone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H10O6
CAS Number:
Molecular Weight:
178.14
Beilstein:
83002
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

form

solid

optical activity

[α]19/D +55°, c = 4 in H2O

mp

187-190 °C (lit.)

storage temp.

2-8°C

SMILES string

OC[C@H](O)[C@H]1OC(=O)[C@@H](O)[C@H]1O

InChI

1S/C6H10O6/c7-1-2(8)5-3(9)4(10)6(11)12-5/h2-5,7-10H,1H2/t2-,3+,4-,5+/m0/s1

InChI key

SXZYCXMUPBBULW-SKNVOMKLSA-N

Application

  • Development and validation of an analysis method: Discusses the use of L-gulonic acid γ-lactone as a matrix effect inhibitor in the validation of a method for pesticide residues by gas chromatography–tandem mass spectrometry (Saegusa, Nomura, Takao, Hamaguchi, 2021).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Beata A Wolucka et al.
The FEBS journal, 273(19), 4435-4445 (2006-09-08)
The last step of the biosynthesis of L-ascorbic acid (vitamin C) in plants and animals is catalyzed by L-gulono-1,4-lactone oxidoreductases, which use both L-gulono-1,4-lactone and L-galactono-1,4-lactone as substrates. L-gulono-1,4-lactone oxidase is missing in scurvy-prone, vitamin C-deficient animals, such as humans
Purification and characterization of L-gulonolactone oxidase from chicken kidney microsomes.
K Kiuchi et al.
Biochemistry, 21(20), 5076-5082 (1982-09-28)
L Braun et al.
FEBS letters, 388(2-3), 173-176 (1996-06-17)
The relationship between glutathione deficiency, glycogen metabolism and ascorbate synthesis was investigated in isolated murine hepatocytes. Glutathione deficiency caused by various agents increased ascorbate synthesis with a stimulation of glycogen breakdown. Increased ascorbate synthesis from UDP-glucose or gulonolactone could not
F Puskás et al.
FEBS letters, 430(3), 293-296 (1998-08-04)
The orientation of gulonolactone oxidase activity was investigated in rat liver microsomes. Ascorbate formation upon gulonolactone addition resulted in higher intravesicular than extravesicular ascorbate concentrations in native microsomal vesicles. The intraluminal ascorbate accumulation could be prevented or the accumulated ascorbate
A Krasnov et al.
Genetic analysis : biomolecular engineering, 15(3-5), 115-119 (1999-12-22)
The reviewed studies addressed the possibility of using gene transfer for correction of L-ascorbic acid biosynthesis and carbohydrate utilization in rainbow trout. Analyses of enzymatic activities in the L-AAB pathway indicated that reasons for the lack of L-AA production can

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service