809799
Nicotinamide-2,6,7-13C3-(pyridyl-15N)
≥98 atom %, ≥98% (CP)
Synonym(s):
Vitamin B3-2,6,7-13C-(pyridyl-15N)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
13C3C3H615NNO
Molecular Weight:
126.10
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.12
Recommended Products
isotopic purity
13C
15N
≥98 atom %
Quality Level
Assay
≥98% (CP)
form
powder
technique(s)
mass spectrometry (MS): suitable
color
white to off-white
mp
128 °C
mass shift
M+4
storage temp.
−20°C
SMILES string
N[13C](C1=CC=[13CH][15N]=[13CH]1)=O
InChI
1S/C6H6N2O/c7-6(9)5-2-1-3-8-4-5/h1-4H,(H2,7,9)/i3+1,4+1,6+1,8+1
InChI key
DFPAKSUCGFBDDF-DNUYGDBSSA-N
General description
Nicotinamide (or niacinamide) is a form of vitamin B3 that can be used in a variety of therapeutic applications. It is found in meat, fish, nuts, and mushrooms, as well as in some vegetables. It is an amide derivative of nicotinic acid and is preferred in the treatment of pellagra, acne and skin cancer.
Nicotinamide-2,6,7-13C3-(pyridyl-15N) is an isotope of vitamin B3 wherein pyridine nitrogen and C-2, C-6, C-7 pyridine carbons are replaced by 15N7 and 13C6 isotopes, respectively.
Nicotinamide-2,6,7-13C3-(pyridyl-15N) is an isotope of vitamin B3 wherein pyridine nitrogen and C-2, C-6, C-7 pyridine carbons are replaced by 15N7 and 13C6 isotopes, respectively.
Application
Nicotinamide-2,6,7-13C3-(pyridyl-15N) can be used as a stable isotope internal standard for accurate data interpretation in specific biological samples.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
302.0 °F - closed cup
Flash Point(C)
150 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
A review of nicotinamide: treatment of skin diseases and potential side effects
Rolfe HM
Journal of Cosmetic Dermatology, 13(4), 324-328 (2014)
Safety of high-dose nicotinamide: a review
Knip M, et al.
Diabetologia, 43(11), 1337-1345 (2000)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service