Skip to Content
Merck
All Photos(2)

Key Documents

792063

Sigma-Aldrich

Copper(I) iodide

anhydrous, 99.995% trace metals basis

Synonym(s):

Cuprous iodide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CuI
CAS Number:
Molecular Weight:
190.45
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
99.995% trace metals basis
grade:
anhydrous
form:
powder
Pricing and availability is not currently available.

grade

anhydrous

Quality Level

vapor pressure

10 mmHg ( 656 °C)

Assay

99.995% trace metals basis

form

powder

impurities

≤55.0 ppm Trace Metal Analysis

mp

605 °C (lit.)

density

5.62 g/mL at 25 °C (lit.)

application(s)

battery manufacturing

SMILES string

[Cu+].[I-]

InChI

1S/Cu.HI/h;1H/q+1;/p-1

InChI key

LSXDOTMGLUJQCM-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Copper Iodide has many applications in organic reactions, including living radical polymerizations (Atom Transfer Radical Polymerization or ATRP)[1] and cross coupling reactions[2]. CuI has also been used in the fabrication of solar cells[3], photoluminescents[4], and nanocrystal synthesis[5].

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1A - STOT RE 1 Oral

Target Organs

Thyroid

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Highly Luminescent CuInS2/ZnS Core/Shell Nanocrystals: Cadmium-Free Quantum Dots for In Vivo Imaging
Li, Liang;
Chemistry of Materials, 21(12), 2422-2429 (2009)
Dawei Ma et al.
Accounts of chemical research, 41(11), 1450-1460 (2008-08-14)
Copper-assisted Ullmann-type coupling reactions are valuable transformations for organic synthesis. Researchers have extensively applied these reactions in both academic and industrial settings. However, two important issues, the high reaction temperatures (normally above 150 degrees C) and the stoichiometric amounts of
Fabrication of an Efficient Solid-State Dye-Sensitized Solar Cell
Meng, Q. B.;
Langmuir, 19(9), 3572-3574 (2003)
Self-assembly of nanometer-scale [Cu24I10L12]14+ cages and ball-shaped Keggin clusters into a (4,12)-connected 3D framework with photoluminescent and electrochemical properties.
Xin-Long Wang et al.
Angewandte Chemie (International ed. in English), 45(44), 7411-7414 (2006-11-07)
Virgil Percec et al.
Journal of the American Chemical Society, 128(43), 14156-14165 (2006-10-26)
Conventional metal-catalyzed organic radical reactions and living radical polymerizations (LRP) performed in nonpolar solvents, including atom-transfer radical polymerization (ATRP), proceed by an inner-sphere electron-transfer mechanism. One catalytic system frequently used in these polymerizations is based on Cu(I)X species and N-containing

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service