Skip to Content
Merck
All Photos(1)

Key Documents

779903

Sigma-Aldrich

Hexamethylphosphoramide

absolute, over molecular sieve (H2O ≤0.03%), ≥98.0% (GC)

Synonym(s):

HMPA, Hexamethylphosphoric acid triamide, Tris(dimethylamino)phosphine oxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)2N]3PO
CAS Number:
Molecular Weight:
179.20
Beilstein:
1099903
EC Number:
MDL number:
UNSPSC Code:
12352111
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

vapor density

6.18 (vs air)

Quality Level

vapor pressure

0.07 mmHg ( 25 °C)

Assay

≥98.0% (GC)

form

liquid

quality

absolute, over molecular sieve (H2O ≤0.03%)

impurities

≤0.03% H2O (coulometric)

refractive index

n20/D 1.459 (lit.)

bp

230-232 °C/740 mmHg (lit.)

mp

7 °C (lit.)

density

1.03 g/mL at 25 °C (lit.)

functional group

phosphoramide

SMILES string

CN(C)P(=O)(N(C)C)N(C)C

InChI

1S/C6H18N3OP/c1-7(2)11(10,8(3)4)9(5)6/h1-6H3

InChI key

GNOIPBMMFNIUFM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Hexamethylphosphoramide can be used:
  • As an additive to facilitate the enantioselective addition of alkyne to aldehydes to generate propargylic alcohols at room temperature.[1]
  • As a catalyst for the reversible-deactivation radical polymerization of methyl methacrylate with an alkyl iodide initiator to generate poly(methyl methacrylate)(PMMA).[2]
  • To facilitate the conversion of tetra-alkyl methanediphosphonates into (Z)-2-aryl-1-(2-cyanoethyl)ethenylphos-phonates.[3]

Hexamethylphosphoramide is a dipolar co-solvent and an effective additive used in reactions like reduction of halides, deoxygenation of sulfones, halide olefin couplings and cleavage of carbon-sulfur bond.[4]

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - Eye Dam. 1 - Muta. 1B - Skin Corr. 1C

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

291.2 °F - closed cup

Flash Point(C)

144 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

?One-pot? synthesis of (Z)-2-aryl-1-(2-cyanoethyl) ethenylphosphonates via hexamethylphosphoramide-promoted sequential transformation
Wang G and Shen Y
Heteroatom Chem., 13(2), 116-119 (2002)
Hexamethylphosphoramide as a highly reactive catalyst for the reversible-deactivation radical polymerization of MMA with an in situ formed alkyl iodide initiator
Wang Y, et al.
Polym. Chem., 8(39), 6073-6085 (2017)
Phenyl- Carbonyl Coupling Reactions Promoted by Samarium Diiodide and Hexamethylphosphoramide
Shiue J, et al.
The Journal of Organic Chemistry, 62(14), 4643-4649 (1997)
Kristopher J Kolonko et al.
The Journal of organic chemistry, 75(18), 6163-6172 (2010-08-26)
A variety of multinuclear NMR techniques, in combination with X-ray diffraction methods, were used to probe the solution structure of α-aryl lithium enolates of bis(4-fluorobenzyl) ketone (1-H), phenyl 4-fluorobenzyl ketone (2-H), and N,N-dimethyl 4-fluorophenylacetamide (3-H) in ethereal solvents and in
Ge Gao et al.
Proceedings of the National Academy of Sciences of the United States of America, 101(15), 5417-5420 (2004-03-18)
It is found that the addition of hexamethylphosphoramide to the solution of an alkyne, Et(2)Zn, and (S)-1,1'-bi-2-naphthol in methylene chloride allows the generation of an alkynylzinc at room temperature and shows highly enantioselective additions to aldehydes. The mild condition for

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service