Skip to Content
Merck
All Photos(2)

Documents

436534

Sigma-Aldrich

N-(Isobutoxymethyl)acrylamide

technical grade

Synonym(s):

N -(Isobutoxymethyl)acrylamide, Isobutoxymethylacrylamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CHCONHCH2OCH2CH(CH3)2
CAS Number:
Molecular Weight:
157.21
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

grade

technical grade

form

viscous liquid

contains

200 ppm monomethyl ether hydroquinone as inhibitor

refractive index

n20/D 1.461 (lit.)

bp

108 °C (lit.)

density

0.97 g/mL at 25 °C (lit.)

SMILES string

CC(C)COCNC(=O)C=C

InChI

1S/C8H15NO2/c1-4-8(10)9-6-11-5-7(2)3/h4,7H,1,5-6H2,2-3H3,(H,9,10)

InChI key

KCTMTGOHHMRJHZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Carc. 1B - Eye Dam. 1 - Muta. 1B - Repr. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 2 - STOT RE 2 Oral

Target Organs

Peripheral nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

174.2 °F - closed cup

Flash Point(C)

79 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sinuo Tan et al.
Journal of chromatography. A, 1625, 461298-461298 (2020-07-28)
This study has examined the batch binding behaviour of different thermo-responsive co-polymer grafted chromatographic materials under different temperature and protein loading conditions. The effect of molecular composition of poly(N-isopropylacrylamide) (PNIPAAm)-based co-polymers on the phase transition properties has been documented. Sixteen

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service