Skip to Content
Merck
All Photos(1)

Key Documents

765767

Sigma-Aldrich

1,1,1-Trifluoro-3-methylbutan-2-one

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H7F3O
CAS Number:
Molecular Weight:
140.10
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.320

bp

52-60 °C/760 mmHg

density

1.080 g/mL at 25 °C

functional group

fluoro
ketone

InChI

1S/C5H7F3O/c1-3(2)4(9)5(6,7)8/h3H,1-2H3

InChI key

NLCCAFQWEKMQLM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-2.2 °F

Flash Point(C)

-19 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service