Skip to Content
Merck
1,1,1-Trifluoro-3-methylbutan-2-one AldrichCPR

1,1,1-Trifluoro-3-methylbutan-2-one

1,1,1-Trifluoro-3-methylbutan-2-one AldrichCPR
Empirical Formula (Hill Notation):
C5H7F3O
CAS Number:
Molecular Weight:
140.10

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing