751162
Propyl methacrylate
contains ~200 ppm MEHQ, 97%
Synonym(s):
n-Propyl methacrylate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C7H12O2
CAS Number:
Molecular Weight:
128.17
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
Quality Level
Assay
97%
form
liquid
contains
~200 ppm MEHQ
refractive index
n20/D 1.419
density
0.898 g/mL at 25 °C
storage temp.
2-8°C
SMILES string
CCCOC(=O)C(C)=C
InChI
1S/C7H12O2/c1-4-5-9-7(8)6(2)3/h2,4-5H2,1,3H3
InChI key
NHARPDSAXCBDDR-UHFFFAOYSA-N
Related Categories
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
96.1 °F
Flash Point(C)
35.6 °C
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Kareem Yusuf et al.
Journal of chromatography. A, 1406, 299-306 (2015-07-05)
A composite zeolitic imidazolate framework-8 (ZIF-8) with a butyl methacrylate-co-ethylene dimethacrylate (BuMA-co-EDMA) monolithic capillary column (33.5cm long×250μm i.d.) was fabricated to enhance the separation efficiency of methacrylate monoliths toward small molecules using conventional low-pressure gas chromatography in comparison with a
Enrique J Carrasco-Correa et al.
Journal of separation science, 38(17), 3028-3037 (2015-06-25)
Polymethacrylimide foams are used as light structural materials in outer-space devices; however, the foam closed cells contain volatile compounds that are outgassed even at low temperatures. These compounds ignite as plasmas under outer-space radiation and the intense radio-frequency fields used
Yun Lei et al.
Journal of pharmaceutical and biomedical analysis, 94, 118-124 (2014-02-25)
A molecularly imprinted polymer coated stir bar (MIP-SB) using dopamine as a template was fabricated. The adsorptive capacity of the MIP-SB was almost 4 times over that of non-imprinted stir bar. The MIP-SB showed good extracting selectivity for dopamine when
Thuy V Nguyen et al.
Biomaterials, 35(22), 5749-5759 (2014-04-15)
Tumor progression is coincident with mechanochemical changes in the extracellular matrix (ECM). We hypothesized that tumor stroma stiffening, alongside a shift in the ECM composition from a basement membrane-like microenvironment toward a dense network of collagen-rich fibers during tumorigenesis, confers
Fangfang Chu et al.
Langmuir : the ACS journal of surfaces and colloids, 31(22), 5992-6000 (2015-01-31)
We study the structure and viscoelastic behavior of 3D plastic crystals of colloidal dumbbells in an oscillatory shear field based on a combination of small-angle neutron scattering experiments under shear (rheo-SANS) and Brownian dynamics computer simulations. Sterically stabilized dumbbell-shaped microgels
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service