Skip to Content
Merck
All Photos(1)

Key Documents

736252

Sigma-Aldrich

Triethyl-d15 phosphate

99 atom % D, 98% (CP)

Synonym(s):

Ethyl-d15 phosphate, TEP-d15

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2D5O)3PO
Molecular Weight:
197.25
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:

isotopic purity

99 atom % D

Assay

98% (CP)

form

liquid

bp

215 °C (lit.)

density

1.160 g/mL at 25 °C

mass shift

M+15

SMILES string

[2H]C([2H])([2H])C([2H])([2H])OP(=O)(OC([2H])([2H])C([2H])([2H])[2H])OC([2H])([2H])C([2H])([2H])[2H]

InChI

1S/C6H15O4P/c1-4-8-11(7,9-5-2)10-6-3/h4-6H2,1-3H3/i1D3,2D3,3D3,4D2,5D2,6D2

InChI key

DQWPFSLDHJDLRL-NLBPYYKNSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lihong Gao et al.
The Science of the total environment, 544, 143-149 (2015-12-15)
The occurrence and distribution of 14 organophosphate (OP) triesters and 5 diesters were investigated in sludge from eight sewage treatment plants (STPs) in Beijing, China, during 2008-2014. Tri(2-ethylhexyl) phosphate (TEHP) and tri-m-cresyl phosphate (TCrP) were the predominant triesters with the
Jinjian Ding et al.
Environmental pollution (Barking, Essex : 1987), 233, 986-991 (2017-10-19)
Foodstuffs may be contaminated by organophosphate esters (OPEs) and become an important source of human exposure since OPEs are ubiquitous in the environment. In the present study, 10 OPEs were analyzed in various food matrices collected from a city in
Xinhong Li et al.
Archives of environmental contamination and toxicology, 78(2), 206-215 (2019-10-28)
Organophosphate flame retardants (OPFRs) are an emerging class of pollutants. In this study, 62 soil samples were collected from Zhejiang Province China, and screened for 8 typical OPFRs. All compounds were detected in soil at 100% detection frequency, except for
Binnian Wei et al.
ACS omega, 4(13), 15364-15372 (2019-10-02)
Emerging chemicals of concern (ECCs), including phthalate plasticizers, flame retardants, and phenolic compounds, are likely present in electronic nicotine delivery system (ENDS) replacement solutions (e-liquids) which are often packaged, stored in, and/or can contact with, plastic, glass, and metal materials.
Nannan Zhao et al.
Journal of agricultural and food chemistry, 68(42), 11694-11701 (2020-09-02)
Animal protein supplement feeds (APFs) are important raw feed materials for livestock. APFs might be susceptible to organophosphate esters (OPEs) but have not been paid attention yet. In the present study, animal-derived (meat meal, feather meal, and blood meal) and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service