Skip to Content
Merck
All Photos(1)

Key Documents

684279

Sigma-Aldrich

2,2,3,3-Tetrafluoropropyl acrylate-1-13C,2,3,3-d3

98 atom % 13C, 98 atom % D, 97% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD2=CD13CO2CH2CF2CHF2
Molecular Weight:
190.12
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:

isotopic purity

98 atom % 13C
98 atom % D

Assay

97% (CP)

form

liquid

mass shift

M+4

SMILES string

[2H]\C([2H])=C(/[2H])[13C](=O)OCC(F)(F)C(F)F

InChI

1S/C6H6F4O2/c1-2-4(11)12-3-6(9,10)5(7)8/h2,5H,1,3H2/i1D2,2D,4+1

InChI key

VHJHZYSXJKREEE-ZPYCZKLBSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Dam. 1 - Flam. Liq. 3 - Muta. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

114.8 °F - closed cup

Flash Point(C)

46 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service