Skip to Content
Merck
All Photos(2)

Key Documents

731528

Sigma-Aldrich

trans-2-Ethoxyvinylboronic acid pinacol ester

95%

Synonym(s):

(E)-2-(2-Ethoxyvinyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, (E)-2-Ethoxyvinylboronic acid pinacol ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H19BO3
CAS Number:
Molecular Weight:
198.07
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

liquid

refractive index

n20/D 1.447

density

0.935 g/mL at 25 °C

functional group

ether

storage temp.

2-8°C

SMILES string

CCO\C=C\B1OC(C)(C)C(C)(C)O1

InChI

1S/C10H19BO3/c1-6-12-8-7-11-13-9(2,3)10(4,5)14-11/h7-8H,6H2,1-5H3/b8-7+

InChI key

MRAYNLYCQPAZJN-BQYQJAHWSA-N

Application

trans-2-Ethoxyvinylboronic acid pinacol ester is a boronic ester commonly used in Suzuki-Miyaura cross-coupling.
This reaction is a key step to synthesize:
  • Azaindole and diazaindoles from chloroamino-N-heterocycles.
  • Doryanine and its derivatives from 2-bromobenzoic acid.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

188.1 °F

Flash Point(C)

86.7 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Total Synthesis of the Illicium-Derived Sesquineolignan Simonsol C.
Nugent, Jeremy et al.
Organic Letters, 18(15), 3798-3801 (2016)
Two-step synthesis of aza-and diazaindoles from chloroamino-N-heterocycles using ethoxyvinylborolane.
Whelligan, Daniel K et al.
The Journal of Organic Chemistry, 75(1), 11-15 (2009)
Efficient and rapid synthesis of N-substituted isoquinolin-1-ones under mild conditions: Facile access to doryanine derivatives
Takwale AD, et al.
Tetrahedron Letters, 60(18), 1259-1261 (2019)
Structure-and reactivity-based development of covalent inhibitors of the activating and gatekeeper mutant forms of the epidermal growth factor receptor (EGFR).
Ward, Richard A et al.
Journal of Medicinal Chemistry, 56(17), 7025-7048 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service