Skip to Content
Merck
All Photos(1)

Key Documents

726508

Sigma-Aldrich

(R)-(+)-Styrene oxide

ChiPros®, produced by BASF, ≥98%

Synonym(s):

(R)-(+)-Phenyloxirane, (R)-Phenylethylene oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H8O
CAS Number:
Molecular Weight:
120.15
Beilstein:
1984
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

grade

produced by BASF

Assay

≥98%
≥98.0% (GC)

form

liquid

optical purity

enantiomeric excess: ≥98.0%

expl. lim.

~22 %

refractive index

n20/D 1.534 (lit.)

bp

89-90 °C/23 mmHg (lit.)

density

1.051 g/mL at 25 °C (lit.)

functional group

ether
phenyl

SMILES string

C1O[C@@H]1c2ccccc2

InChI

1S/C8H8O/c1-2-4-7(5-3-1)8-6-9-8/h1-5,8H,6H2/t8-/m0/s1

InChI key

AWMVMTVKBNGEAK-QMMMGPOBSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

ChiPros is a registered trademark of BASF SE

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Carc. 1B - Eye Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

176.0 °F - closed cup

Flash Point(C)

80 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shuangjun Lin et al.
Organic letters, 12(17), 3816-3819 (2010-08-14)
Neocarzinostatin (1) biosynthesis is proposed to involve a vicinal diol intermediate. It is reported that NcsF2, one of two epoxide hydrolases encoded by the NCS gene cluster, catalyzes regiospecific addition of H(2)O to C-2 of both (R)- and (S)-styrene oxides
Jung-Hee Woo et al.
Biotechnology letters, 35(4), 599-606 (2012-12-18)
A novel epoxide hydrolase (EHase) from polycyclic aromatic hydrocarbon (PAH)-degrading bacteria was identified and characterized. EHase activity was identified in four strains of PAH-degrading bacteria isolated from commercial gasoline and oil-contaminated sediment based on their growth on styrene oxide and
Gary P Carlson
Drug and chemical toxicology, 34(4), 440-444 (2011-07-14)
Styrene is known to be hepatotoxic and pneumotoxic in rodents, and these adverse effects are related to its metabolism. Mice deficient in the enzymes responsible for both the activation and detoxification of styrene are useful in examining this relationship more
Alessandra Lattanzi et al.
Chirality, 22 Suppl 1, E130-E135 (2010-11-03)
Density Functional Theory (DFT) calculations of optical rotation (OR) and vibrational circular dichroism (VCD) have been used to assign the absolute configuration (AC) of a recently prepared (3-phenyloxirane-2,2-diyl)bis(phenylmethanone), 3, by asymmetric epoxidation of the corresponding 2-arylidene-1,3-diketone. The experimental OR at
Laurent Gaté et al.
Toxicology letters, 211(3), 211-219 (2012-04-18)
The cytogenetic alterations in leukocytes and the increased risk for leukemia, lymphoma, or all lymphohematopoietic cancer observed in workers occupationally exposed to styrene have been associated with its hepatic metabolisation into styrene-7,8-oxide, an epoxide which can induce DNA damages. However

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service