726427
3,5-Dimethylbenzyl bromide
≥97.0% (GC)
Synonym(s):
α-Bromomesitylene, 1-(Bromomethyl)-3,5-dimethylbenzene
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C9H11Br
CAS Number:
Molecular Weight:
199.09
Beilstein:
1857343
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
≥97.0% (GC)
form
powder
mp
37-39 °C (lit.)
functional group
bromo
SMILES string
Cc1cc(C)cc(CBr)c1
InChI
1S/C9H11Br/c1-7-3-8(2)5-9(4-7)6-10/h3-5H,6H2,1-2H3
InChI key
QXDHXCVJGBTQMK-UHFFFAOYSA-N
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service