Skip to Content
Merck
All Photos(2)

Key Documents

692352

Sigma-Aldrich

Magnesium bis(hexamethyldisilazide)

97%

Synonym(s):

Bis(hexamethyldisilazido)magnesium, Mg(HMDS)2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H36MgN2Si4
CAS Number:
Molecular Weight:
345.07
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

97%

form

solid

mp

121-124 °C

SMILES string

C[Si](C)(C)N([Mg]N([Si](C)(C)C)[Si](C)(C)C)[Si](C)(C)C

InChI

1S/2C6H18NSi2.Mg/c2*1-8(2,3)7-9(4,5)6;/h2*1-6H3;/q2*-1;+2

InChI key

WYPTZCBYSQFOQS-UHFFFAOYSA-N

Application

Magnesium bis(hexamethyldisilazide) is an essential precursor in the synthesis of wide range of diketiminate ligand complexes for rac-lactide polymerization.[1][2][3] It can also be used in the synthesis of ItBu:Mg[N(SiMe3)2]2, where ItBu = (1,3-di-tert-butylimidazol-2-ylidene), a catalyst for the guanylation reaction of addition of amine to carbodiimide.[4]

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Highly active magnesium initiators for ring-opening polymerization of rac-lactide.
Wang L and Ma H
Macromolecules, 43(16), 6535-6537 (2010)
Catalytic C?N bond formation in guanylation reaction by N-heterocyclic carbene supported magnesium (II) and zinc (II) amide complexes.
Baishya A, et al.
Journal of Organometallic Chemistry, 769, 112-118 (2014)
Stereoselectivity switch between zinc and magnesium initiators in the polymerization of rac-lactide: different coordination chemistry, different stereocontrol mechanisms.
Wang H, et al.
Macromolecules, 47(22), 7750-7764 (2014)
Lactide polymerization catalyzed by Mg and Zn diketiminate complexes with flexible ligand frameworks.
Whitehorne T J, et al.
Dalton Transactions, 43(17), 6339-6352 (2014)
Dong Chen et al.
Nanoscale, 11(48), 23173-23181 (2019-11-30)
CoSe2 materials with different nanostructures are used as pseudocapacitive Mg-storage cathodes, which exhibit fast solid-state Mg2+ ions diffusion kinetics. In this work, CoSe2 with different nanostructures including hollow microspheres (H-CoSe2), nano-polyhedra (P-CoSe2) and nanorods (R-CoSe2) are fabricated by using facile

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service