Skip to Content
Merck
All Photos(3)

Key Documents

324620

Sigma-Aldrich

Lithium bis(trimethylsilyl)amide

97%

Synonym(s):

Hexamethyldisilazane lithium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)3Si]2NLi
CAS Number:
Molecular Weight:
167.33
EC Number:
MDL number:
UNSPSC Code:
12352111
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

pH

(12.4 at 23,7 °C)

bp

110-115 °C (at 1.33 hPa)

mp

70-71 °C

density

0.860 g/mL at 25 °C (lit.)

SMILES string

[Li]N([Si](C)(C)C)[Si](C)(C)C

InChI

1S/C6H18NSi2.Li/c1-8(2,3)7-9(4,5)6;/h1-6H3;/q-1;+1

InChI key

YNESATAKKCNGOF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Lithium bis(trimethylsilyl)amide is a non-nucleophilic strong Brønsted base, which is generally soluble in most of the nonpolar organic solvents. It is most commonly employed in organic reactions.

Application

Base employed in generating enolates for the preparation of lactone precursors, pyranones, and cyclohexanes.
Used to catalyze the addition of phosphine P-H bonds to carbodiimides leading to phosphaguanidines. Also used in a novel three-step synthesis of disubstituted 1,2,5-thiadiazoles.

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Sol. 1 - Self-heat. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 2

Flash Point(F)

62.6 °F - closed cup

Flash Point(C)

17 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron, 50, 9061-9061 (1994)
Ana I Ojeda-Amador et al.
Inorganic chemistry, 55(11), 5719-5728 (2016-05-14)
Reacting cesium fluoride with an equimolar n-hexane solution of lithium bis(trimethylsilyl)amide (LiHMDS) allows the isolation of CsHMDS (1) in 80% yield (after sublimation). This preparative route to 1 negates the need for pyrophoric Cs metal or organocesium reagents in its
Tetrahedron Letters, 47, 8285-8285 (2006)
Lithium Bis(trimethylsilyl)amide and Tris(trimethylsilyl)amine
Inorganic Syntheses (1966)
Synlett, 507-507 (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service