673935
Chlorodiphenylsilane
technical grade, 90%
Synonym(s):
Diphenylchlorosilane
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
(C6H5)2SiHCl
CAS Number:
Molecular Weight:
218.75
Beilstein:
2937164
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
grade
technical grade
Quality Level
Assay
90%
refractive index
n20/D 1.578 (lit.)
bp
143 °C/10 mmHg (lit.)
density
1.118 g/mL at 25 °C (lit.)
SMILES string
[H][Si](Cl)(c1ccccc1)c2ccccc2
InChI
1S/C12H11ClSi/c13-14(11-7-3-1-4-8-11)12-9-5-2-6-10-12/h1-10,14H
InChI key
VTMOWGGHUAZESJ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Chlorodiphenylsilane is an organosilicon compound that can be used as a protecting group for alcohols and amines. It is also used as a reagent in cross-coupling reactions. In addition, it is used as a precursor in the synthesis of various silicon-containing compounds.
Application
Chlorodiphenylsilane can be used as a reactant for the selective mono- and di-allylation and allenylation of chlorosilanes in the presence of indium/base to synthesize mono- or di-allylated or allenylation products. It also acts as a responsive coupling agent for functionalizing silica surfaces.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Skin Corr. 1B
Supplementary Hazards
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 1
Flash Point(F)
221.0 °F
Flash Point(C)
105 °C
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service