Skip to Content
Merck
All Photos(1)

Key Documents

113379

Sigma-Aldrich

Chloro(dimethyl)phenylsilane

98%

Synonym(s):

DMPSCl, Dimethylphenylchlorosilane, Dimethylphenylsilylchloride, Phenyldimethylchlorosilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5Si(CH3)2Cl
CAS Number:
Molecular Weight:
170.71
Beilstein:
606292
EC Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

refractive index

n20/D 1.509 (lit.)

density

1.017 g/mL at 25 °C (lit.)

SMILES string

C[Si](C)(Cl)c1ccccc1

InChI

1S/C8H11ClSi/c1-10(2,9)8-6-4-3-5-7-8/h3-7H,1-2H3

InChI key

KWYZNESIGBQHJK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Used in a synthesis of enantioenriched allenylsilanes via an ortho-ester Claisen rearrangement of chiral, silylpropargylic alcohols.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

143.6 °F - closed cup

Flash Point(C)

62 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ryan A Brawn et al.
Organic letters, 9(14), 2689-2692 (2007-06-15)
A convenient procedure for the synthesis of highly enantioenriched allenylsilanes by Johnson orthoester Claisen rearrangement of 1-silyl propargylic alcohols is described. Allenylsilanes are then used as carbon nucleophiles in three-component, Lewis acid mediated additions to in situ generated oxonium ions
Yixuan Han et al.
The Journal of chemical physics, 153(4), 044902-044902 (2020-08-06)
Density changes in thin polymer films have long been considered as a possible explanation for shifts in the thickness-dependent glass transition temperature Tg(h) in such nanoconfined systems, given that the glass transition is fundamentally associated with packing frustration during material
Carlton P Folster et al.
Dalton transactions (Cambridge, England : 2003), 49(45), 16125-16132 (2020-01-30)
The synthesis of a chloro-functionalized six-membered cyclosilazane (Si5N) is reported. Subsequent reductive polymerizations yielded low molecular weight polysilazanes. 1H and 29Si NMR characterization suggest the identity of the reducing metal influences the polysilazane structure. Optical characterization is consistent with extended
M Okamoto
Yakugaku zasshi : Journal of the Pharmaceutical Society of Japan, 112(6), 409-413 (1992-06-01)
The retention behavior and selectivity of 30 kinds of phenyl-modified porous glasses and silicas, prepared from solutions of phenyldimethylchlorosilane, diphenylmethylchlorosilane or triphenylchlorosilane in xylene, were studied by high-performance liquid chromatography using carbamazepine and diphenylhydantoin as model compounds. From the elemental

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service