Skip to Content
Merck
All Photos(1)

Key Documents

671479

Sigma-Aldrich

cataCXium® A

95%

Synonym(s):

Di(1-adamantyl)-n-butylphosphine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H39P
CAS Number:
Molecular Weight:
358.54
Beilstein:
8726448
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

reaction suitability

reagent type: ligand
reaction type: Arylations

reagent type: ligand
reaction type: Buchwald-Hartwig Cross Coupling Reaction

reagent type: ligand
reaction type: C-C Bond Formation

reagent type: ligand
reaction type: Cross Couplings

reagent type: ligand
reaction type: Heck Reaction

reagent type: ligand
reaction type: Sonogashira Coupling

reagent type: ligand
reaction type: Suzuki-Miyaura Coupling

functional group

phosphine

SMILES string

CCCCP([C@]12C[C@H]3C[C@H](C[C@H](C3)C1)C2)[C@@]45C[C@@H]6C[C@@H](C[C@@H](C6)C4)C5

InChI

1S/C24H39P/c1-2-3-4-25(23-11-17-5-18(12-23)7-19(6-17)13-23)24-14-20-8-21(15-24)10-22(9-20)16-24/h17-22H,2-16H2,1H3/t17-,18+,19-,20-,21+,22-,23-,24-

InChI key

HTJWUNNIRKDDIV-FECFBMJZSA-N

General description

Sold in collaboration with Solvias AG

cataCXium® A is commonly used as a catalyst in cross-coupling reactions, such as Suzuki and Sonogashira reactions.

Application

cataCXium® A or di-adamantylalkylphosphine is a bulky and electron-rich phosphine ligand that is highly effective for palladium catalyzed cross-coupling reactions such as Heck and Suzuki coupling, Buchwald-Hartwig amination of aryl chlorides, and α-arylation reactions of ketones.

Other applications:
  • palladium-catalyzed carbonylation of aryl and heteroaryl halides
  • palladium-catalyzed synthesis of (hetero)aromatic nitriles
  • palladium-catalyzed aminocarbonylation of aryl halides

Features and Benefits

  • Mild reaction condition
  • Low catalyst loading
  • High yield and turn over number

Legal Information

US7148176
cataCXium is a registered trademark of Umicore AG & Co. KG

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Selective Palladium-Catalyzed Aminocarbonylation of Aryl Halides with CO and Ammonia
Wu, Xiao-Feng and Neumann, Helfried and Beller, Matthias
Chemistry?A European Journal , 16(32), 9750-9753 (2010)
Palladium-catalyzed carbonylation reactions of aryl halides and related compounds
Brennfuhrer A, et al.
Angewandte Chemie (International Edition in English), 48(23), 4114-4133 (2009)
Efficient carbonylation of aryl and heteroaryl bromides using a palladium/diadamantylbutylphosphine catalyst
Neumann H, et al.
Advanced Synthesis & Catalysis, 348(10-11), 1255-1261 (2006)
A general and efficient method for the formylation of aryl and heteroaryl bromides
Klaus S, et al.
Angewandte Chemie (International Edition in English), 45(1), 154-158 (2006)
Efficient carbonylation of aryl and heteroaryl bromides using a palladium/diadamantylbutylphosphine catalyst
Neumann H, et al.
advanced synthesis and catalysis, 348(10-11), 1255-1261 (2006)

Articles

cataCXium® ligands facilitate cross-coupling reactions, essential in polymer science, fine chemicals, and pharmaceutical industries.

cataCXium® ligands facilitate cross-coupling reactions, essential in polymer science, fine chemicals, and pharmaceutical industries.

cataCXium® ligands facilitate cross-coupling reactions, essential in polymer science, fine chemicals, and pharmaceutical industries.

cataCXium® ligands facilitate cross-coupling reactions, essential in polymer science, fine chemicals, and pharmaceutical industries.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service