669083
2,2,2-Trifluoroethyl formate
95%
Synonym(s):
TFEF
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CF3CH2OC(O)H
CAS Number:
Molecular Weight:
128.05
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
95%
refractive index
n20/D <1.001
density
1.317 g/mL at 25 °C
functional group
fluoro
SMILES string
[H]C(=O)OCC(F)(F)F
InChI
1S/C3H3F3O2/c4-3(5,6)1-8-2-7/h2H,1H2
InChI key
CAFROQYMUICGNO-UHFFFAOYSA-N
Related Categories
Application
Reagent for the formylation of alcohols and amines. Hydroxylamines provide high yields of the N-formylated product.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
45.0 °F - closed cup
Flash Point(C)
7.2 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
David R Hill et al.
Organic letters, 4(1), 111-113 (2002-01-05)
[reaction: see text] Treatment of a variety of alcohols, amines, and N-hydroxylamines with 2,2,2-trifluoroethyl formate gave the corresponding formylated adducts in high yields.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service