Skip to Content
Merck
All Photos(3)

Key Documents

662534

Sigma-Aldrich

4-Mercaptobenzoic acid

technical grade, 90%

Synonym(s):

4-Carboxybenzenethiol, 4-Carboxythiophenol, 4-Mercaptobenzoic acid, p-Carboxybenzenethiol, p-Mercaptobenzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HSC6H4CO2H
CAS Number:
Molecular Weight:
154.19
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Quality Level

Assay

90%

form

solid

mp

215-224 °C (lit.)

functional group

carboxylic acid

SMILES string

OC(=O)c1ccc(S)cc1

InChI

1S/C7H6O2S/c8-7(9)5-1-3-6(10)4-2-5/h1-4,10H,(H,8,9)

InChI key

LMJXSOYPAOSIPZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Mercaptobenzoic acid (4-MBA), also known as 4-Carboxythiophenol or p-Mercaptobenzoic acid, is an aromatic compound used in organic synthesis. 4-MBA promotes the formation of disulfide bonds through its reactive thiol (-SH) group. The sulfur atom in the thiol group is highly nucleophilic, allowing it to attack electrophiles.  facilitates the formation of diverse thiol-containing compounds.  Additionally, 4-MBA serves as a ligand to form coordination complexes and is utilized for surface functionalization, particularly in creating self-assembled monolayers (SAMs) on metal surfaces.

Application

  • SERS substrate for food adulteration detection: 4-Mercaptobenzoic acid was employed in the layer-by-layer biopolymer assembly for the in situ fabrication of AuNP plasmonic paper, serving as a SERS substrate useful in detecting food adulteration (Viriyakitpattana et al., 2024).
  • Development of magneto-plasmonic nanoparticles: Research highlighted the use of 4-Mercaptobenzoic acid in the multiwavelength SERS of magneto-plasmonic nanoparticles, obtained through combined laser ablation and solvothermal methods, showing versatility in nanoparticle synthesis (Talaikis et al., 2023).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chitosan-4-mercaptobenzoic acid: synthesis and characterization of a novel thiolated chitosan
Millotti et al.
Journal of Materials Chemistry, 20, 2432-2440 (2010)
Coordination chemistry of 3-and 4-mercaptobenzoate ligands: versatile hydrogen-bonding isomers of the thiosalicylate (2-mercaptobenzoate) ligand
Tiekink et al.
Coordination Chemistry Reviews, 341, 19-52 (2017)
A theoretical study of 4-Mercaptobenzoic acid assembled on Ag for surface-enhanced raman scattering applications
Mhlanga et al.
Materials Today Communications, 26, 101698-101698 (2021)
Electrochemical and infrared spectroscopy studies of 4-mercaptobenzoic acid SAMs on gold surfaces
Rosendahl et al.
Electrochimica Acta, 53, 6759-6767 (2008)
Aleksandra Jaworska et al.
The Analyst, 140(7), 2321-2329 (2014-12-09)
The intracellular pH plays an important role in various cellular processes. In this work, we describe a method for monitoring of the intracellular pH in endothelial cells by using surface enhanced Raman spectroscopy (SERS) and 4-mercaptobenzoic acid (MBA) anchored to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service