Skip to Content
Merck
All Photos(1)

Key Documents

524018

Sigma-Aldrich

4-Mercaptophenylboronic acid

90%

Synonym(s):

(4-Sulfanylphenyl)boronic acid, B-(4-Mercaptophenyl)boronic acid, Dihydroxy(4-mercaptophenyl)borane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HSC6H4B(OH)2
CAS Number:
Molecular Weight:
153.99
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

90%

form

solid

mp

>230 °C (lit.)

SMILES string

OB(O)c1ccc(S)cc1

InChI

1S/C6H7BO2S/c8-7(9)5-1-3-6(10)4-2-5/h1-4,8-10H

InChI key

AUVSUPMVIZXUOG-UHFFFAOYSA-N

General description

4-Mercaptophenylboronic acid is a stable boronic acid that can be used as a reagent in the Pd-catalyzed Suzuki-Miyaura cross-coupling reactions to prepare useful building blocks via C-C bond formation.

Application

4-Mercaptophenylboronic acid can be used to functionalize Fe3O4-C-Au magnetic microspheres to synthesize core−shell structure Fe3O4-C-Au magnetic microspheres for the selective enrichment of glycoproteins and glycopeptides.

Other Notes

may contain bis-4,4′-thiophenylboronic

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shuangshuang Li et al.
Physical chemistry chemical physics : PCCP, 17(27), 17638-17645 (2015-06-18)
Molecular recognition based on specific intermolecular interactions is essential for the design of sensors with high selectivity. Herein, we report the surface-enhanced Raman scattering (SERS) behaviour of 4-mercaptophenyl boronic acid (MPBA) on self-assembled silver nanoparticles and its interaction with d-glucose.
Siyue Gao et al.
Journal of microbiological methods, 147, 69-75 (2018-03-20)
The existence of pathogenic bacteria in drinking water has been a threat to the safety of human well-being. Traditional methods to detect bacteria are standard plate counts or rapid methods such as ELISA and PCR. However, those methods can be
Runmei Li et al.
Materials (Basel, Switzerland), 12(11) (2019-06-06)
The difference in gold nanoparticle (AuNPs) aggregation caused by different mixing orders of AuNPs, 4-mercaptophenylboronic acid (4-MPBA), and hydrogen peroxide (H2O2) has been scarcely reported. We have found that the color change of a ((4-MPBA + AuNPs) + H2O2) mixture
Xiao Wang et al.
Talanta, 201, 119-125 (2019-05-28)
Glycated hemoglobin (HbA1c) represents the average glucose level over the past three months and has been considered as the most important biomarker for the diagnosis of Type Ⅱ diabetes (T2D). Herein, a label-free and quantitative electrochemical biosensor based on 4-mercaptophenylboronic
Zhao Yu et al.
Mikrochimica acta, 185(7), 330-330 (2018-06-20)
A surface-enhanced Raman scattering (SERS) method has been developed to determine the concentration of trichloroethylene (TCE) in environmental water. Au-core/Ag-shell nanoparticles containing 4-mercaptophenylboronic acid (4-MPBA) between the core and shell are used as the SERS substrate. 4-MPBA serves as an

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service