Skip to Content
Merck
All Photos(1)

Documents

648639

Sigma-Aldrich

2,4-Pentanedione, polymer-bound

100-200 mesh particle size, extent of labeling: 2.5-3.5 mmol/g O loading, 2 % cross-linked with divinylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352116
NACRES:
NA.22

Quality Level

crosslinking

2 % cross-linked with divinylbenzene

reaction suitability

reaction type: solution phase peptide synthesis
reactivity: nucleophile reactive

extent of labeling

2.5-3.5 mmol/g O loading

particle size

100-200 mesh

storage temp.

2-8°C

Related Categories

Application

Scavenges hydrazines and pimary amines.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Uwe Schoun, Josef Messinger, Nuria Merayo, Grzegorz Juszkiewicz, Andreas Kirschning
Synlett, 07, 983-986 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service