Skip to Content
Merck
All Photos(1)

Key Documents

641979

Sigma-Aldrich

Ethyl 4-iodobenzoate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H9IO2
CAS Number:
Molecular Weight:
276.07
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.5880 (lit.)

density

1.641 g/mL at 25 °C (lit.)

functional group

ester
iodo

SMILES string

CCOC(=O)c1ccc(I)cc1

InChI

1S/C9H9IO2/c1-2-12-9(11)7-3-5-8(10)6-4-7/h3-6H,2H2,1H3

InChI key

YCBJOQUNPLTBGG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Ethyl 4-iodobenzoate can be used in the synthesis of biphenyl derivatives via Negishi cross coupling with the intermediates of the type ArMg(tmp)·2LiCl (tmp = trimethylolpropane). It can also be used as a starting material in the synthesis of p-carbomethoxyphenylmagnesium bromide, a polyfunctional organometallic reagent that can be used as building block in the synthesis of complex target molecules.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

"Direct Magnesiation of Polyfunctionalized Arenes and Heteroarenes Using (tmp) 2Mg? 2 LiCl"
Clososki.CG, et al.
Angewandte Chemie (Weinheim an der Bergstrasse, Germany), 46(40), 7681-7684 (2007)
"Preparation and reactions of functionalized arylmagnesium reagents"
Jensen EA, et al.
Synthesis, 2002(04), 0565-0569 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service